Merck
  • Cadmium exposure and clinical cardiovascular disease: a systematic review.
Cadmium exposure and clinical cardiovascular disease: a systematic review.
Current atherosclerosis reports (2013-08-21)
Maria Tellez-Plaza, Miranda R Jones, Alejandro Dominguez-Lucas, Eliseo Guallar, Ana Navas-Acien
ABSTRACT
Mounting evidence supports that cadmium, a toxic metal found in tobacco, air and food, is a cardiovascular risk factor. Our objective was to conduct a systematic review of epidemiologic studies evaluating the association between cadmium exposure and cardiovascular disease. Twelve studies were identified. Overall, the pooled relative risks (95% confidence interval) for cardiovascular disease, coronary heart disease, stroke, and peripheral arterial disease were: 1.36 (95% CI: 1.11, 1.66), 1.30 (95% CI: 1.12, 1.52), 1.18 (95% CI: 0.86, 1.59), and 1.49 (95% CI: 1.15, 1.92), respectively. The pooled relative risks for cardiovascular disease in men, women and never smokers were 1.29 (1.12, 1.48), 1.20 (0.92, 1.56) and 1.27 (0.97, 1.67), respectively. Together with experimental evidence, our review supports the association between cadmium exposure and cardiovascular disease, especially for coronary heart disease. The number of studies with stroke, heart failure (HF) and peripheral arterial disease (PAD) endpoints was small. More studies, especially studies evaluating incident endpoints, are needed.
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Cadmium, wire reel, 0.1m, diameter 2.0mm, hard, 99.99+%
Cadmium, wire reel, 0.5m, diameter 0.25mm, hard, 99.99+%
Cadmium, foil, 10mm disks, thickness 0.25mm, as rolled, 99.95%
Cadmium, wire reel, 0.1m, diameter 1.0mm, hard, 99.999%
Cadmium, foil, 0.5m coil, thickness 0.5mm, as rolled, 100%
Cadmium, foil, 25mm disks, thickness 0.125mm, as rolled, 99.95%
Cadmium, foil, 100x100mm, thickness 0.125mm, as rolled, 99.95%
Cadmium, foil, 25mm disks, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, 25mm disks, thickness 0.125mm, as rolled, 99.99+%
Cadmium, foil, 25x25mm, thickness 0.5mm, as rolled, 99.999%
Cadmium, foil, 15mm disks, thickness 0.125mm, as rolled, 99.95%
Cadmium, foil, 100x100mm, thickness 0.125mm, as rolled, 99.99+%
Cadmium, wire reel, 0.2m, diameter 0.5mm, hard, 99.95+%
Cadmium, foil, 15mm disks, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 10mm disks, thickness 0.015mm, 99.7+%
Cadmium, foil, 300x300mm, thickness 2.0mm, as rolled, 99.99%
Cadmium, foil, 50x200mm, thickness 1.0mm, as rolled, 99.999%
Cadmium, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99%
Cadmium, wire reel, 0.5m, diameter 0.70mm, as drawn, 99.99+%
Cadmium, foil, 10mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, wire reel, 0.5m, diameter 0.5mm, hard, 99.95+%
Cadmium, foil, 50x50mm, thickness 1.0mm, as rolled, 99.95%
Cadmium, foil, 200x200mm, thickness 1.0mm, as rolled, 99.99%
Cadmium, foil, 150x150mm, thickness 0.25mm, as rolled, 99.95%
Cadmium, wire reel, 0.2m, diameter 1.0mm, hard, 99.999%
Cadmium, foil, 10mm disks, thickness 0.25mm, as rolled, 99.9%