Merck
  • MauG, a diheme enzyme that catalyzes tryptophan tryptophylquinone biosynthesis by remote catalysis.
MauG, a diheme enzyme that catalyzes tryptophan tryptophylquinone biosynthesis by remote catalysis.
Archives of biochemistry and biophysics (2013-10-23)
Sooim Shin, Victor L Davidson
ABSTRACT
MauG contains two c-type hemes with atypical physical and catalytic properties. While most c-type cytochromes function simply as electron transfer mediators, MauG catalyzes the completion of tryptophan tryptophylquinone (TTQ)(1) biosynthesis within a precursor protein of methylamine dehydrogenase. This posttranslational modification is a six-electron oxidation that requires crosslinking of two Trp residues, oxygenation of a Trp residue and oxidation of the resulting quinol to TTQ. These reactions proceed via a bis-Fe(IV) state in which one heme is present as Fe(IV)O and the other is Fe(IV) with axial heme ligands provided by His and Tyr side chains. Catalysis does not involve direct contact between the protein substrate and either heme of MauG. Instead it is accomplished by remote catalysis using a hole hopping mechanism of electron transfer in which Trp residues of MauG are reversibly oxidized. In this process, long range electron transfer is coupled to the radical mediated chemical reactions that are required for TTQ biosynthesis.
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Tryptophan, European Pharmacopoeia (EP) Reference Standard
Supelco
Tryptophan, Pharmaceutical Secondary Standard; Certified Reference Material