Merck
Saccharin.
Diabetes care (1989-01-01)
S A Miller, V P Frattali
ABSTRACT
Saccharin, a nonnutritive sweetener discovered in 1879, has been the subject of controversy concerning its effect on public health on several occasions during this century. Over this period, the substance has come to be regarded as a useful commodity in the dietary management of diabetes mellitus. We review the historical and scientific background on the subject and propose a new approach in making public-health decisions on unique foods that serve a special dietary purpose.
Product Description
USP
Saccharin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Saccharin, ≥99%
Sigma-Aldrich
Saccharin, ≥98%
Supelco
Saccharin, Pharmaceutical Secondary Standard; Certified Reference Material
Saccharin, European Pharmacopoeia (EP) Reference Standard
Supelco
Mettler-Toledo Calibration substance ME 51143091, Saccharin, traceable to primary standards (LGC)