Merck
  • Nandrolone decanoate: pharmacological properties and therapeutic use in osteoporosis.
Nandrolone decanoate: pharmacological properties and therapeutic use in osteoporosis.
Clinical rheumatology (1995-09-01)
P Geusens
ABSTRACT
The therapeutic profile on bone of nandrolone decanoate is that of inhibitor of bone resorption with temporary increase in bone formation, followed by an absence of suppression of bone formation, indicating uncoupling of bone resorption and formation. This results is an increase in bone mineral content at the proximal and distal radius, and in some patients at the lumbar spine. Furthermore, nandrolone decanoate increases calcium balance and muscle mass, diminishes vertebral pain and increases the mobility of the spine. Virilization occurred in around 50% of the patients (mainly hoarseness and/or hirsutism), and 9% of the patients dropped out because of this reason. A dose of 50 mg every 3 to 4 weeks is indicated in the treatment of osteoporosis in women, especially when they have low muscle mass, associated debilitating disease, and in patients with corticosteroid induced osteoporosis. It should only be prescribed after the age of 65 to 75 years to minimize the occurrence of clinical adverse effects and to increase its tolerability, which is higher in this group. Although some effects are reported on fracture rate, insufficient prospective data are available.
Sigma-Aldrich
19-Nortestosterone, ≥99.0% (HPLC)
Supelco
Nandrolone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Nandrolone decanoate for peak identification, European Pharmacopoeia (EP) Reference Standard
Nandrolone decanoate, European Pharmacopoeia (EP) Reference Standard
Nandrolone decanoate for system suitability, European Pharmacopoeia (EP) Reference Standard