Merck
  • Preparation and evaluation of monolithic poly(N-vinylcarbazole-co-1,4-divinylbenzene) capillary columns for the separation of small molecules.
Preparation and evaluation of monolithic poly(N-vinylcarbazole-co-1,4-divinylbenzene) capillary columns for the separation of small molecules.
Analytical and bioanalytical chemistry (2014-07-25)
Rainer Koeck, Martin Fischnaller, Rania Bakry, Richard Tessadri, Guenther K Bonn
ABSTRACT
Short-term polymerization or the so-called low-conversion polymerization was applied for the preparation of N-vinylcarbazole (NVC) and 1,4-divinylbenzene (DVB) monolithic capillary columns. The synthesis was carried out by thermally initiated free radical copolymerization under the influence of inert micro- (toluene) and macroporogen (1-decanol) and α,α'-azoisobutyronitrile (AIBN) as radical initiator. The morphological and porous properties were studied by scanning electron microscopy (SEM), nitrogen adsorption, and mercury intrusion porosimetry (MIP). The copolymerization process was studied by monomer conversion measurements. This approach led to increased porosity and specific surface area. A specific surface area above 400 m(2)/g of the monolith and a distinct bimodal pore size distribution were obtained. The chromatographic performance was determined in terms of theoretical plate heights and number of theoretical plates. The lowest plate height value was found to be 3.9 μm (corresponding to ≈256,000 plates per meter) applying methylparaben utilizing an 80 mm × 0.2 mm i.d. monolithic capillary. The developed NVC/DVB monolithic supports showed high separation efficiency towards small molecules, which was exemplified applying reversed-phase (RP) separation of alkylbenzenes, beta-blockers, flavanoids, parabens, and phenones. The loading capacity was analyzed for isocratic separation of seven alkylbenzenes and was found to be up to 77 ng total mass of alkylbenzenes. Furthermore, a long-term stability test of 1,000 consecutive runs was performed and resulted in a maximum variance of 0.97, 0.85, and 0.16 % RSD for resolution, peak width at half height, and retention times, respectively. The material was proven to have a high permeability of 1.11E-14 m(2), applying water as a mobile phase.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
1-Decanol, 98%
Sigma-Aldrich
1-Decanol, ≥98%
Supelco
1-Decanol, analytical standard
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, 98%
Sigma-Aldrich
1-Decanol, ≥98%, FCC, FG
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)