Merck
  • Rapid and simultaneous determination of sulfonate ester genotoxic impurities in drug substance by liquid chromatography coupled to tandem mass spectrometry: comparison of different ionization modes.
Rapid and simultaneous determination of sulfonate ester genotoxic impurities in drug substance by liquid chromatography coupled to tandem mass spectrometry: comparison of different ionization modes.
Journal of chromatography. A (2014-07-06)
Tian Guo, Yuanyuan Shi, Li Zheng, Feng Feng, Feng Zheng, Wenyuan Liu
ABSTRACT
Two ionization techniques for liquid chromatography-mass spectrometry (LC-MS) determination of sulfonate ester potentially genotoxic impurities (PGIs) were evaluated. Twelve PGIs including methyl, ethyl, propyl and isopropyl esters of methanesulfonate, benzenesulfonate and p-toluenesulfonate were studied in this research. Electrospray ionization (ESI) and atmospheric pressure chemical ionization (APCI) sources were compared in terms of performance and quality parameters for detection of the twelve PGIs. Their mass spectra obtained by APCI and ESI were very different in both fragment ions and relative abundances. In APCI negative ion mode the twelve sulfonate esters showed their stable precursor ions of [M-alkyl](-), which readily yielded product ions of [M-alkyl-CH3](-) (for aliphatic sulfonate esters) or [M-alkyl-SO2](-) (for aromatic sulfonate esters) with collision-induced dissociation (CID) applied; and working in selected reaction monitoring (SRM) mode has allowed limits of detection to be decreased. In the case of ESI ionization, these compounds showed their precursor ions [M+H](+), but their abundance was easily competed by formation of ions [M+NH4](+) and/or [M+Na](+), which led to poor analytical sensitivity and reproducibility. Although mobile phase additives could enhance the responses of adduct ions like [M+NH4](+) and [M+Na](+), no improvement was obtained when using SRM mode. Twelve sulfonate esters were systematically compared and APCI was shown to be a better ionization technique for rapid and sensitive determination of these PGIs. Performance of the developed approach for rapid determination of 12 PGIs was also evaluated. Quality parameters were established and good precision (relative standard deviations <8%) and very low limits (2-4ng/mL) of detection were obtained, mainly when using APCI in SRM mode.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ultrapure Acetonitrile
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Methyl p-toluenesulfonate, 98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%