Merck
An easy access to 7-methyl-2-naphthalenecarbonitrile.
Chemical & pharmaceutical bulletin (2005-04-02)
Hiroyuki Koshio, Tsukasa Ishihara, Hiroyoshi Yamada, Fukushi Hirayama, Yuzo Matsumoto, Isao Yanagisawa
ABSTRACT
A practical and cost-effective procedure has been developed for the synthesis of 7-methyl-2-naphthalenecarbonitrile, the precursor of the anticoagulant agents YM-60828 or YM-96765. This new route generates the key intermediate in only two steps from readily available 3-cyanopropionaldehyde dimethyl acetal and m-tolualdehyde, without requiring chromatographic purification. The synthesis involves condensation of the cyano derivative with the aldehyde and subsequent cyclodehydration.
Product Description
Supelco
Discovery® Cyano Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm