Merck
  • Risk assessment for the Italian population of acetaldehyde in alcoholic and non-alcoholic beverages.
Risk assessment for the Italian population of acetaldehyde in alcoholic and non-alcoholic beverages.
Food chemistry (2014-02-13)
Viviana Paiano, Giancarlo Bianchi, Enrico Davoli, Eva Negri, Roberto Fanelli, Elena Fattore
ABSTRACT
Acetaldehyde is a naturally-occurring carcinogenic compound, present in different food items, especially in alcoholic beverages. The aims of this study were to measure acetaldehyde concentration in different beverages consumed in Italy and to estimate the potential cancer risk. The analytical procedure was based on headspace solid-phase microextraction (SPME) and gas chromatography-mass spectrometry (GC-MS), using the isotopic dilution method. The margin of exposure (MOE) approach of the European Food Safety Authority (EFSA) was used for risk characterisation. The highest concentrations (median, min-max) were detected in grappa samples (499, 23.4-1850mg/l), followed by fruit-based liqueurs and spirits (62.0, 5.23-483mg/l) and wine (68.0, 18.1-477mg/l); the lowest were detected in gin (0.91, 0.78-1.90mg/l). The lowest MOE was estimated for high wine consumers (69). These results suggest that regulatory measures and consumer guidance may be necessary for acetaldehyde in beverages.
Sigma-Aldrich
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Supelco
Acetaldehyde, PESTANAL®, analytical standard
Sigma-Aldrich
Acetaldehyde, natural, FG
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF