Merck
Staphylococcal biofilm formation as affected by type acidulant.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica (2013-12-11)
Antonia Nostro, Luigina Cellini, Giovanna Ginestra, Manuela D'Arrigo, Mara di Giulio, Andreana Marino, Anna Rita Blanco, Angelo Favaloro, Giuseppe Bisignano
ABSTRACT
Staphylococcal growth and biofilm formation in culture medium where pH was lowered with weak organic (acetic and lactic) or strong inorganic (hydrochloric) acids were studied. The effects were evaluated by biomass measurements, cell-surface hydrophobicity, scanning electron microscopy (SEM), and confocal laser scanning microscopy (CLSM). The results demonstrated that the inhibition was related to type of acidulant and pH value. At pH 5.0, the antibacterial effect was more pronounced in the presence of acetic acid (58-60% growth reduction) compared with that in the presence of lactic (7-16% growth reduction) and hydrochloric acids (23-24% reduction). The biofilm biomass of Staphylococcus aureus and Staphylococcus epidermidis was reduced by 92, 85, 63, and 93, 87, 81% after exposition to acetic, lactic, and hydrochloric acids, respectively. Increasing the pH from 5.0 to 6.0 resulted in a noticeable reduction in the effectiveness of acids. A minor cells hydrophobic character was also documented. The SEM and CLSM revealed a poorly structured and thinner biofilm compared with the dense and multilayered control. Acidic environment could have important implications for food-processing system to prevent bacterial colonization and control biofilm formation. The findings of this study lead to consider the rational use of the type of acid to achieve acidic environments.
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Lactic acid, natural, ≥85%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Lactic acid, 88%, FCC
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Supelco
Lactic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%
Supelco
Hydrogen chloride - 1-butanol solution, ~3 M in 1-butanol, derivatization grade (GC derivatization), LiChropur