Merck
  • The impact of roller compaction and tablet compression on physicomechanical properties of pharmaceutical excipients.
The impact of roller compaction and tablet compression on physicomechanical properties of pharmaceutical excipients.
Pharmaceutical development and technology (2013-08-15)
Raman Mahadevan Iyer, Shridhar Hegde, James Dinunzio, Dharmendra Singhal, Waseem Malick
ABSTRACT
Material properties play a significant role in pharmaceutical processing. The impact of roller compaction (RC) and tablet compression on solid fraction (SF), tensile strength (TS) and flexural modulus (FM) of Avicel DG [co-processed excipient with 75% microcrystalline cellulose (MCC) and 25% anhydrous dibasic calcium phosphate (DCPA)], lactose and 1:1 Mixture of the two was studied. Materials were roller compacted at different force and roller type and compressed into tablets over a range of compression pressures (CP). SF, TS and FM were determined for ribbons and tablets. Roller force was a significant variable affecting SF while roller type was not. Both SF and TS of tablets increased with CP with Avicel DG exhibiting greater TS than that of 1:1 Mixture while tablets of lactose had the lowest TS. The TS of tablets decreased exponentially with tablet porosity. Ribbon of Avicel DG had higher TS and lower SF than lactose and greater reworkability. This is attributed to plastic deformation of MCC resulting in high degree of bonding and fragmentation of DCPA that fills the void spaces during tablet compression. The lack of significant increase in SF and low tablet TS for lactose upon compression is likely due to its brittle fragmentation and some elastic recovery as shown by the high FM.
Product Description
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
α-Cellulose, powder
Sigma-Aldrich
Dibasic Calcium Phosphate, meets USP testing specifications
Sigma-Aldrich
Calcium phosphate dibasic, 98.0-105.0%
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
USP
Anhydrous lactose, United States Pharmacopeia (USP) Reference Standard
Supelco
Cellulose, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Sigma-Aldrich
Cellulose, colloidal, microcrystalline
Sigma-Aldrich
Cellulose, microcrystalline, powder
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Sigma-Aldrich
Lactose, tested according to Ph. Eur.
Supelco
Cellulose, acid washed, from spruce, suitable for column chromatography
Supelco
Cellulose, powder, suitable for column chromatography
Supelco
Cellulose, acid washed, powder, suitable for column chromatography
Supelco
Cellulose, DS-0, powder, suitable for thin layer chromatography (TLC)
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm
Sigma-Aldrich
Cellulose, fibers, (medium)
Lactose (anhydrous), European Pharmacopoeia (EP) Reference Standard
Supelco
Lactose, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material