Merck
  • Chlorhexidine-impregnated dressing for prevention of catheter-related bloodstream infection: a meta-analysis*.
Chlorhexidine-impregnated dressing for prevention of catheter-related bloodstream infection: a meta-analysis*.
Critical care medicine (2014-03-29)
Nasia Safdar, John C O'Horo, Aiman Ghufran, Allison Bearden, Maria Eugenia Didier, Dan Chateau, Dennis G Maki
ABSTRACT
To assess the efficacy of a chlorhexidine-impregnated dressing for prevention of central venous catheter-related colonization and catheter-related bloodstream infection using meta-analysis. Multiple computerized database searches supplemented by manual searches including relevant conference proceedings. Randomized controlled trials evaluating the efficacy of a chlorhexidine-impregnated dressing compared with conventional dressings for prevention of catheter colonization and catheter-related bloodstream infection. Data were extracted on patient and catheter characteristics and outcomes. Nine randomized controlled trials met the inclusion criteria. Use of a chlorhexidine-impregnated dressing resulted in a reduced prevalence of catheter-related bloodstream infection (random effects relative risk, 0.60; 95% CI, 0.41-0.88, p = 0.009). The prevalence of catheter colonization was also markedly reduced in the chlorhexidine-impregnated dressing group (random effects relative risk, 0.52; 95% CI, 0.43-0.64; p < 0.001). There was significant benefit for prevention of catheter colonization and catheter-related bloodstream infection, including arterial catheters used for hemodynamic monitoring. Other than in low birth weight infants, adverse effects were rare and minor. Our analysis shows that a chlorhexidine-impregnated dressing is beneficial in preventing catheter colonization and, more importantly, catheter-related bloodstream infection and warrants routine use in patients at high risk of catheter-related bloodstream infection and central venous catheter or arterial catheter colonization.
Product Description
Supelco
Chlorhexidine, Pharmaceutical Secondary Standard; Certified Reference Material
Chlorhexidine dihydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Chlorhexidine dihydrochloride, ≥98%
Sigma-Aldrich
Chlorhexidine, ≥99.5%
Chlorhexidine, European Pharmacopoeia (EP) Reference Standard