Merck
  • The impact of topical corticosteroid use before diagnosis on the outcome of Acanthamoeba keratitis.
The impact of topical corticosteroid use before diagnosis on the outcome of Acanthamoeba keratitis.
Ophthalmology (2014-03-19)
Dana Robaei, Nicole Carnt, Darwin C Minassian, John K G Dart
ABSTRACT
To examine the impact of topical corticosteroid use before the diagnosis of Acanthamoeba keratitis (AK) on final visual outcomes and to determine the prognostic factors predicting poorer outcomes. Cohort study. A total of 209 eyes of 196 patients with retrievable medical records, diagnosed with AK at Moorfields Eye Hospital, London, between January 1991 and April 2012. One eye was randomly excluded from analysis in the 13 cases of bilateral AK. Patient demographic, initial clinical examination findings, and management details were collected. The outcomes of patients treated with topical corticosteroids before diagnosis of AK were compared with those not treated with topical corticosteroids before diagnosis. A multivariable logistic model, optimized for prior corticosteroid use, was used to derive the odds ratios (ORs) of a suboptimal visual outcome. Suboptimal visual outcome was defined as final visual acuity (VA) ≤ 20/80, corneal perforation, or need for keratoplasty. Acanthamoeba keratitis was diagnosed on microbiological culture in 94 eyes (48.0%), on histopathologic examination in 27 eyes (13.8%), on confocal microscopy in 38 eyes (19.4%), and on the basis of a typical clinical course and response to treatment in 37 eyes (18.9%). Final VA and prior corticosteroid use data were available for 174 eyes (88.8%). In multivariable analysis, corticosteroid use before diagnosis was associated with suboptimal visual outcome (OR, 3.90; 95% confidence interval [CI], 1.78-8.55), as were disease stage 3 at presentation (OR, 5.62; 95% CI, 1.59-19.80) and older age (60+ years) at diagnosis (OR, 8.97; 95% CI, 2.13-37.79). Corticosteroid use before diagnosis of AK is highly predictive of a poorer visual outcome. This is largely due to the initial misdiagnosis of AK as herpetic keratitis. It is important to include AK in the differential diagnosis of keratitis in all contact lens users with keratitis, particularly before making a diagnosis of herpes keratitis and before the use of topical corticosteroids in the therapy of any indolent keratitis.
Sigma-Aldrich
Chlorhexidine, ≥99.5%
Sigma-Aldrich
Chlorhexidine dihydrochloride, ≥98%
Chlorhexidine, European Pharmacopoeia (EP) Reference Standard
Supelco
Chlorhexidine, Pharmaceutical Secondary Standard; Certified Reference Material
Chlorhexidine dihydrochloride, European Pharmacopoeia (EP) Reference Standard