Merck
  • Overexpression of Iris. lactea var. chinensis metallothionein llMT2a enhances cadmium tolerance in Arabidopsis thaliana.
Overexpression of Iris. lactea var. chinensis metallothionein llMT2a enhances cadmium tolerance in Arabidopsis thaliana.
Ecotoxicology and environmental safety (2014-05-02)
Chun-Sun Gu, Liang-qin Liu, Yan-Hai Zhao, Yan-ming Deng, Xu-dong Zhu, Su-Zhen Huang
ABSTRACT
Metallothioneins (MTs) are cysteine-rich, low molecular weight, heavy metal-binding protein molecules. Here, a full-length cDNA homologue of MT2a (type 2 metallothionein) was isolated from the cadmium-tolerant species Iris. lactea var. chinensis (I. lactea var. chinensis). Expression of IlMT2a in I. lactea var. chinensis roots and leaves was up-regulated in response to cadmium stress. When the gene was constitutively expressed in Arabidopsis thaliana (A. thaliana), root length of transgenic lines was longer than that of wild-type under 50μM or 100μM cadmium stress. However, there was no difference of cadmium absorption between wild-type and trangenic lines. Histochemical staining by 3,3-diaminobenzidine (DAB) and nitroblue tetrazoliu (NBT) clearly demonstrated that transgenic lines accumulated remarkably less H2O2 and O2(-) than wild-type. Together, IlMT2a may be a promising gene for the cadmium tolerance improvement.
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Cadmium, wire reel, 0.1m, diameter 2.0mm, hard, 99.99+%
Cadmium, wire reel, 0.1m, diameter 1.2mm, as drawn, 99.99+%
Cadmium, wire reel, 0.1m, diameter 0.70mm, as drawn, 99.99+%
Cadmium, foil, not light tested, 100x100mm, thickness 0.01mm, 99.7+%
Cadmium, foil, 8mm disks, thickness 0.25mm, as rolled, 99.95%
Cadmium, foil, 6mm disks, thickness 0.5mm, as rolled, 99.95%
Cadmium, foil, 6mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Cadmium, foil, not light tested, 50x50mm, thickness 0.005mm, 99.7+%
Cadmium, foil, not light tested, 25x25mm, thickness 0.01mm, 99.7+%
Cadmium, wire reel, 1m, diameter 1.2mm, as drawn, 99.99+%
Cadmium, wire reel, 2m, diameter 0.5mm, hard, 99.95+%
Cadmium, foil, 4mm disks, thickness 0.125mm, as rolled, 99.95%
Cadmium, foil, 10mm disks, thickness 0.125mm, as rolled, 99.99+%
Cadmium, foil, 25mm disks, thickness 0.5mm, as rolled, 99.95%
Cadmium, wire reel, 2m, diameter 0.5mm, hard, 99.99+%
Cadmium, foil, 100x100mm, thickness 0.125mm, as rolled, 99.95%
Cadmium, foil, 4mm disks, thickness 0.125mm, as rolled, 99.99+%
Cadmium, wire reel, 5m, diameter 0.5mm, hard, 99.95+%
Cadmium, wire reel, 5m, diameter 0.5mm, hard, 99.99+%
Cadmium, foil, 50x200mm, thickness 1.0mm, as rolled, 99.999%
Cadmium, wire reel, 1m, diameter 0.5mm, hard, 99.95+%
Cadmium, foil, 2m coil, thickness 0.5mm, as rolled, 100%
Cadmium, foil, 25mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, wire reel, 0.1m, diameter 0.125mm, as drawn, 99.9%
Cadmium, wire reel, 2m, diameter 0.125mm, as drawn, 99.9%