Merck
  • Mitochondrial dysfunction in early life resulted from perinatal bisphenol A exposure contributes to hepatic steatosis in rat offspring.
Mitochondrial dysfunction in early life resulted from perinatal bisphenol A exposure contributes to hepatic steatosis in rat offspring.
Toxicology letters (2014-04-29)
Ying Jiang, Wei Xia, Yingshuang Zhu, Xiaocui Li, Danqi Wang, Juan Liu, Huailong Chang, Gengqi Li, Bing Xu, Xi Chen, Yuanyuan Li, Shunqing Xu
ABSTRACT
An emerging literature suggests that bisphenol A (BPA), a widespread endocrine disrupting chemical, when exposure occurs in early life, may increase the risk of metabolic syndrome. In this study, we investigated the hypothesis that perinatal exposure to BPA predisposed offspring to fatty liver disease: the hepatic manifestation of metabolic syndrome, and its possible mechanism. Pregnant Wistar rats were administered with BPA (40μg/kg/day) or vehicle during gestation and lactation. Liver histology, biochemical analysis, transcriptome, and mitochondrial function were examined in male offspring at postnatal 3, 15 and 26 weeks. At 3 weeks of age, abnormal liver morphology and function were not observed in the BPA-exposed offspring, but a decrease in mitochondrial respiratory complex (MRC) activity (I and III) and significant changes in gene expression involved in mitochondrial fatty acid metabolism were observed compared with controls. At 15 weeks, micro-vesicular steatosis in liver, up-regulated genes involved in lipogenesis pathways, increased ROS generation and Cytc release were observed in the BPA-exposed offspring. Then, extensive fatty accumulation in liver and elevated serum ALT were observed in BPA-exposed offspring at 26 weeks. In the longitudinal observation, hepatic mitochondrial function including MRC activity, ATP production, ROS generation and mitochondrial membrane potential were progressively worsened in the BPA-exposed offspring. Perinatal BPA exposure contributes to the development of hepatic steatosis in the offspring of rats, which may be mediated through impaired hepatic mitochondrial function and up-regulated hepatic lipid metabolism.
Sigma-Aldrich
ProteoMass Cytochrome c MALDI-MS Standard, vial of 10 nmol, (M+H+) 12,361.96 Da by calculation
Sigma-Aldrich
Cytochrome c from Saccharomyces cerevisiae, ≥85% based on Mol. Wt. 12,588 basis
Supelco
Bisphenol A, ≥99%
Sigma-Aldrich
Bisphenol A, 97%
Sigma-Aldrich
Cytochrome c from equine heart, BioReagent, suitable for GFC marker
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% (SDS-PAGE)
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% based on Mol. Wt. 12,384 basis
Sigma-Aldrich
Cytochrome c from pigeon breast muscle, ≥95% based on Mol. Wt. 12,173 basis
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis, powder, suitable for mammalian cell culture
Supelco
Bisphenol A, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Cytochrome c from equine heart, BioUltra, ≥99% (SDS-PAGE), powder, suitable for mammalian cell culture