Merck
  • Removal of nitrite from aqueous solution using sugarcane bagasse and wheat straw.
Removal of nitrite from aqueous solution using sugarcane bagasse and wheat straw.
Bulletin of environmental contamination and toxicology (2014-05-23)
Dereje Diriba, Ahmed Hussen, Vegi Maheswara Rao
ABSTRACT
Sugarcane bagasse and wheat straw were applied for the removal of nitrite ions from water samples. Batch experiments were conducted to establish optimum pH (5), initial nitrite concentration (5 mg/L), adsorbent dose (3 mg/L) and contact time (90 min). Under the optimized conditions, raw sugarcane bagasse was found to be a more effective (removal efficiency 90 %) adsorbent in removing nitrite ions than wheat straw (removal efficiency 63 %). Adsorption isotherms and kinetic parameters were also studied. The correlation coefficient values for Langmuir and Freundlich isotherm models were 0.9625 and 0.9590, respectively. The results showed that the adsorption of nitrite fairly fits both Langmuir and Freundlich adsorption isotherms for both adsorbents. The kinetics of the adsorption process follows the pseudo second-order kinetic model.
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
α-Cellulose, powder
Supelco
Cellulose, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Sigma-Aldrich
Cellulose, colloidal, microcrystalline
Sigma-Aldrich
Cellulose, microcrystalline, powder
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Supelco
Cellulose, acid washed, from spruce, suitable for column chromatography
Supelco
Cellulose, powder, suitable for column chromatography
Supelco
Cellulose, acid washed, powder, suitable for column chromatography
Supelco
Cellulose, DS-0, powder, suitable for thin layer chromatography (TLC)
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm
Sigma-Aldrich
Cellulose, fibers, (medium)