Merck
  • Influence of the selective EDTA derivative phenyldiaminetetraacetic acid on the speciation and extraction of heavy metals from a contaminated soil.
Influence of the selective EDTA derivative phenyldiaminetetraacetic acid on the speciation and extraction of heavy metals from a contaminated soil.
Chemosphere (2014-05-31)
Tao Zhang, Hang Wei, Xiu-Hong Yang, Bing Xia, Jun-Min Liu, Cheng-Yong Su, Rong-Liang Qiu
ABSTRACT
The development of more selective chelators for the washing of heavy metal contaminated soil is desirable in order to avoid excessive dissolution of soil minerals. Speciation and mobility of Cu, Zn, Pb, and Ni in a contaminated soil washed with phenyldiaminetetraacetic acid (PDTA), a derivative of EDTA, were investigated by batch leaching test using a range of soil washing conditions followed by sequential extraction. With appropriate washing conditions, PDTA significantly enhanced extraction of Cu from the contaminated soil. The primary mechanisms of Cu extraction by PDTA were complexation-promoted dissolution of soil Cu and increased dissolution of soil organic matter (SOM). PDTA showed high selectivity for Cu(II) over soil component cations (Ca(II), Mg(II), Fe(III), Mn(II), Al(III)), especially at lower liquid-to-soil ratios under PDTA deficiency, thus avoiding unwanted dissolution of soil minerals during the soil washing process which can degrade soil structure and interfere with future land use. PDTA-enhanced soil washing increased the exchangeable fractions of Cu, Zn, and Pb and decreased their residual fractions, compared to their levels in unwashed soil.
Sigma-Aldrich
Copper, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, powder (spheroidal), 10-25 μm, 98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Copper, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
Copper, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
Copper, shot, −3-+14 mesh, 99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, Molecular Biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sodium calcium edetate, European Pharmacopoeia (EP) Reference Standard
Copper, IRMM®, certified reference material, 0.1 mm foil
Copper (O), BCR®, certified reference material, rod
Copper (S, P), BCR®, certified reference material, chips
Sigma-Aldrich
Copper, powder, 99.99% trace metals basis
Copper, IRMM®, certified reference material, 0.5 mm wire
Copper, rod, 50mm, diameter 16mm, as drawn, 99.99+%
Copper - O.F.H.C., foil, 10mm disks, thickness 0.010mm, hard, 99.95+%
Copper - O.F.H.C., foil, 1m coil, thickness 0.025mm, hard, 99.95+%
Copper - O.F.H.C., foil, 6mm disks, thickness 0.05mm, hard, 99.95+%
Copper - O.F.H.C., foil, 50mm disks, thickness 0.006mm, hard, 99.95+%
Copper - O.F.H.C., foil, 2m coil, thickness 0.05mm, coil width 300mm, half hard, 99.95+%
Copper - O.F.H.C., foil, 4mm disks, thickness 0.7mm, half hard, 99.95+%