Merck
  • Effects of chemical and thermal stress on acetylcholinesterase activity in the brain of the bank vole, Myodes glareolus.
Effects of chemical and thermal stress on acetylcholinesterase activity in the brain of the bank vole, Myodes glareolus.
Ecotoxicology and environmental safety (2014-05-24)
Renata Świergosz-Kowalewska, Patrycja Molenda, Anna Halota
ABSTRACT
One of the most important issues in ecotoxicology is better understanding the effects of interactions between chemical pollutants and physical environmental factors on animals. To fill this knowledge gap, changes in the activity of acetylcholinesterase (AChE) in the brain samples of bank voles Myodes (Clethrionomys) glareolus due to temperature effects, and two chemical stressors were studied in a full factorial laboratory experiment (27 treatments). The experiment was divided into three phases: acclimatisation (3 days), intoxication (42 days) and elimination (21 days). During the intoxication phase, animals were orally exposed to different concentrations of either nickel (0, 300 or 800 mg Ni/kg food), chlorpyrifos (CPF) (0, 50 or 350 mg CPF/kg food) or a mixture of both chemicals. During the acclimatisation and elimination phases, the bank voles were given uncontaminated food. The experiment was conducted at three different temperatures (10, 20 or 30 °C), and a 12 h:12 h light:dark regime. The animals were sacrificed at 0, 5, 10, 20, 42, 49 and 63 days after the beginning of the intoxication, and brain samples were obtained for chemical analysis. The nickel accumulation in the brain depended on the level of nickel exposure and on interactions between the temperature and other factors. Nickel exhibited no effect on AChE activity. In contrast, AChE was drastically inhibited by chlorpyrifos and low temperature, but interactions between all factors significantly influenced the enzyme activity during the elimination phase of the experiment. High mortality was observed in the groups exposed to high concentrations of nickel and chlorpyrifos.
Product Description
Nickel, foil, 0.5m coil, thickness 0.0075mm, 99.9%
Nickel, foil, 4mm disks, thickness 0.009mm, 99.95%
Nickel, foil, 10mm disks, thickness 0.003mm, 99.95%
Nickel, foil, not light tested, 100x100mm, thickness 0.002mm, 99.95%
Nickel, foil, not light tested, 25x25mm, thickness 0.001mm, 99.95%
Nickel, foil, 0.5m coil, thickness 0.15mm, annealed, 99%
Nickel, wire reel, 100m, diameter 0.5mm, as drawn, 99%
Nickel, tube, 1000mm, outside diameter 0.8mm, inside diameter 0.64mm, wall thickness 0.08mm, as drawn, 99.5%
Nickel, foil, 15mm disks, thickness 0.5mm, as rolled, 99.99+%
Nickel, rod, 100mm, diameter 9.5mm, 99%
Nickel, foil, 1m coil, thickness 0.1mm, annealed, 99%
Nickel, tube, 100mm, outside diameter 0.75mm, inside diameter 0.59mm, wall thickness 0.08mm, hard, 99.5%
Nickel, foil, 1m coil, thickness 0.025mm, 99.9%
Nickel, foil, 15mm disks, thickness 0.25mm, as rolled, 99.99+%
Nickel, rod, 50mm, diameter 10.0mm, 99.99+%
Nickel, rod, 100mm, diameter 12.0mm, 99%
Nickel, foil, 1m coil, thickness 0.5mm, annealed, 99.98%
Nickel, rod, 100mm, diameter 6.35mm, 99%
Nickel, foil, 1m coil, thickness 0.15mm, coil width 12mm, annealed, 99%
Nickel, foil, 15mm disks, thickness 0.025mm, as rolled, 100%
Nickel, rod, 100mm, diameter 12.7mm, 99.98%
Nickel, rod, 50mm, diameter 40mm, 99%
Nickel, rod, 500mm, diameter 12.0mm, 99%
Nickel, foil, 10mm disks, thickness 0.3mm, as rolled, 100%
Nickel, tube, 200mm, outside diameter 0.8mm, inside diameter 0.64mm, wall thickness 0.08mm, as drawn, 99.5%
Nickel, rod, 500mm, diameter 3.3mm, 99.99+%
Nickel, tube, 100mm, outside diameter 0.8mm, inside diameter 0.5mm, wall thickness 0.15mm, as drawn, 99.5%
Nickel, wire reel, 200m, diameter 0.25mm, hard, 99.98%
Nickel, foil, 15mm disks, thickness 0.3mm, annealed, 99%
Nickel, rod, 50mm, diameter 25.4mm, 99.98%