Merck
  • Effects of cadmium exposure on the gill proteome of Cottus gobio: modulatory effects of prior thermal acclimation.
Effects of cadmium exposure on the gill proteome of Cottus gobio: modulatory effects of prior thermal acclimation.
Aquatic toxicology (Amsterdam, Netherlands) (2014-05-31)
Jennifer Dorts, Patrick Kestemont, Marie-Laetitia Thézenas, Martine Raes, Frédéric Silvestre
ABSTRACT
Temperature and trace metals are common environmental stressors, and their importance is increasing due to global climate change and anthropogenic pollution. The aim of the present study was to investigate whether acclimation to elevated temperature affects the response of the European bullhead (Cottus gobio) to subsequent cadmium (Cd) exposure by using enzymatic and proteomic approaches. Fish acclimated to 15 (standard temperature), 18 or 21 °C for 28 days were exposed to 1mg Cd/L for 4 days at the respective acclimation temperature. First, exposure to Cd significantly decreased the activity of the lactate dehydrogenase (LDH) in gills of fish acclimated to 15 or 18 °C. However, an acclimation to 21 °C suppressed the inhibitory effect of Cd. Second, using a proteomic analysis by 2D-DIGE, we observed that thermal acclimation was the first parameter affecting the protein expression profile in gills of C. gobio, while subsequent Cd exposure seemed to attenuate this temperature effect. Moreover, our results showed opposite effects of these two environmental stressors at protein expression level. From the 52 protein spots displaying significant interaction effects of temperature and Cd exposure, a total of 28 different proteins were identified using nano LC-MS/MS and the Peptide and Protein Prophet algorithms of Scaffold software. The identified differentially expressed proteins can be categorized into diverse functional classes, related to protein turnover, folding and chaperoning, metabolic process, ion transport, cell signaling and cytoskeleton. Within a same functional class, we further reported that several proteins displayed reverse responses following sequential exposure to heat and Cd. This work provides insights into the molecular pathways potentially involved in heat acclimation process and the interactive effects of temperature and Cd stress in ectothermic vertebrates.
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Cadmium, foil, not light tested, 25x25mm, thickness 0.01mm, 99.7+%
Cadmium, tube, 500mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cadmium, foil, not light tested, 50x50mm, thickness 0.005mm, 99.7+%
Cadmium, foil, light tested, 25x25mm, thickness 0.005mm, 99.7+%
Cadmium, foil, 6mm disks, thickness 0.5mm, as rolled, 99.95%
Cadmium, foil, 8mm disks, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 6mm disks, thickness 0.015mm, 99.7+%
Cadmium, rod, 50mm, diameter 40mm, 99.9%
Cadmium, foil, 6mm disks, thickness 0.25mm, as rolled, 99.99+%
Cadmium, wire reel, 0.1m, diameter 0.25mm, hard, 99.99+%
Cadmium, foil, not light tested, 50x50mm, thickness 0.015mm, 99.7+%
Cadmium, foil, 6mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, foil, light tested, 25x25mm, thickness 0.015mm, 99.7+%
Cadmium, foil, 4mm disks, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 4mm disks, thickness 0.25mm, as rolled, 99.99+%
Cadmium, foil, 8mm disks, thickness 0.25mm, as rolled, 99.95%
Cadmium, foil, 6mm disks, thickness 0.01mm, 99.7+%
Cadmium, foil, not light tested, 50x50mm, thickness 0.002mm, 99.7+%
Cadmium, tube, 1000mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cadmium, rod, 100mm, diameter 5.0mm, as drawn, 99.99+%
Cadmium, foil, 6mm disks, thickness 1.0mm, as rolled, 99.95%
Cadmium, rod, 200mm, diameter 5.0mm, as drawn, 99.99+%
Cadmium, foil, not light tested, 100x100mm, thickness 0.015mm, 99.7+%
Cadmium, foil, 8mm disks, thickness 0.05mm, as rolled, 99.99+%
Cadmium, rod, 300mm, diameter 10.0mm, as drawn, 99.99+%
Cadmium, foil, light tested, 50x50mm, thickness 0.025mm, 99.7+%