Merck
  • PmMGST3, a novel microsomal glutathione S-transferase gene in the dinoflagellate Prorocentrum minimum, is a potential biomarker of oxidative stress.
PmMGST3, a novel microsomal glutathione S-transferase gene in the dinoflagellate Prorocentrum minimum, is a potential biomarker of oxidative stress.
Gene (2014-05-27)
Ruoyu Guo, Vinitha Ebenezer, Jang-Seu Ki
ABSTRACT
In this study, we evaluated a novel microsomal glutathione S-transferase3 (MGST3) gene from the dinoflagellate Prorocentrum minimum, and examined its expression pattern in response to copper-and nickel-induced stresses. The full length of PmMGST3 was 732 bp, ranging from the dinoflagellate splice leader (DinoSL) sequence to the poly (A) tail, covering a 441-bp ORF, 97-bp 5'UTR, and 194-bp 3'UTR. The PmMGST3 was up-regulated by metals, including copper and nickel. The highest up-regulation levels of the PmMGST3 were found under 0.1 mg/L copper and 0.5 mg/L nickel treatment, respectively. In addition, the PmMGST3 was gradually up-regulated by 0.1 mg/L copper with increasing exposure time. Furthermore, ROS production and reduced GSH was measured in the copper treated cells. A significant increased ROS production and reduced GSH were found in the copper treated cells. These results suggest that PmMGST3 may be related to defense mechanisms associated with oxidative stress in dinoflagellates.
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Copper, tube, 500mm, outside diameter 3.0mm, inside diameter 1.2mm, wall thickness 0.9mm, as drawn, 99.9%
Nickel, foil, 50mm disks, thickness 0.005mm, as rolled, 99.9%
Copper - O.F.H.C., foil, 5m coil, thickness 0.1mm, half hard, 99.95+%
Copper - O.F.H.C., foil, 1m coil, thickness 0.25mm, half hard, 99.95+%
Copper - O.F.H.C., foil, 25mm disks, thickness 0.025mm, hard, 99.95+%
Copper - O.F.H.C., foil, 10m coil, thickness 0.1mm, hard, 99.95+%
Copper, wire reel, 25m, diameter 0.75mm, as drawn, 99.98+%
Copper - O.F.H.C., foil, 10mm disks, thickness 0.1mm, hard, 99.95+%
Nickel, foil, 5m coil, thickness 0.05mm, coil width 2mm, annealed, 99%
Copper - O.F.H.C., foil, 10m coil, thickness 0.125mm, hard, 99.95+%
Copper - O.F.H.C., foil, 15mm disks, thickness 0.125mm, hard, 99.95+%
Copper - O.F.H.C., foil, 25mm disks, thickness 0.7mm, half hard, 99.95+%
Copper, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.999%
Copper - O.F.H.C., foil, 2m coil, thickness 0.20mm, hard, 99.95+%
Nickel, foil, 5m coil, thickness 0.0125mm, 99.9%
Copper, tube, 500mm, outside diameter 4.0mm, inside diameter 3.2mm, wall thickness 0.4mm, as drawn, 99.9%
Copper - O.F.H.C., foil, 10mm disks, thickness 0.05mm, hard, 99.95+%
Copper - O.F.H.C., foil, 50mm disks, thickness 0.5mm, half hard, 99.95+%