Merck
  • Do bacterial cellulose membranes have potential in drug-delivery systems?
Do bacterial cellulose membranes have potential in drug-delivery systems?
Expert opinion on drug delivery (2014-05-23)
Armando J D Silvestre, Carmen S R Freire, Carlos P Neto
ABSTRACT
Bacterial cellulose (BC) is an extremely pure form of cellulose, which, due to its unique properties, such as high purity, water-holding capacity, three-dimensional nanofibrilar network, mechanical strength, biodegradability and biocompatibility, shows a high potential as nanomaterial in a wide range of high-tech domains including biomedical applications, and most notably in controlled drug-delivery systems. This appraisal is intended to cover the major characteristics of BC, followed by the key aspects of BC production both in static and agitated conditions, and a glance of the major applications of BC, giving some emphasis to biomedical applications. Finally, a detailed discussion of the different applications of BC in controlled drug-delivery systems will be put forward, with focus on topical and oral drug-delivery systems, using either native BC or composite materials thereof. The limited number of studies carried out so far demonstrated that BC, or materials prepared from it, are interesting materials for drug-delivery systems. There is, however, a large field of systematic research ahead to develop new and more selectively responsive materials and eventually to conjugate them with other biomedical applications of BC under development.
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
α-Cellulose, powder
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Sigma-Aldrich
Cellulose, fibers, (medium)
Supelco
Cellulose, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Supelco
Cellulose, acid washed, powder, suitable for column chromatography
Sigma-Aldrich
Cellulose, microcrystalline, powder
Supelco
Cellulose, powder, suitable for column chromatography
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm
Sigma-Aldrich
Cellulose, colloidal, microcrystalline
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Supelco
Cellulose, acid washed, from spruce, suitable for column chromatography
Supelco
Cellulose, DS-0, powder, suitable for thin layer chromatography (TLC)