Merck
  • Design, synthesis, and biological evaluations of tumor-targeting dual-warhead conjugates for a taxoid-camptothecin combination chemotherapy.
Design, synthesis, and biological evaluations of tumor-targeting dual-warhead conjugates for a taxoid-camptothecin combination chemotherapy.
Journal of medicinal chemistry (2014-06-06)
Jacob G Vineberg, Edison S Zuniga, Anushree Kamath, Ying-Jen Chen, Joshua D Seitz, Iwao Ojima
ABSTRACT
Novel tumor-targeting dual-warhead conjugates, 2 (DW-1) and 3 (DW-2), which consist of a next-generation taxoid, 1 (SB-T-1214), and camptothecin as two warheads, self-immolative disulfide linkers for drug release, biotin as the tumor-targeting moiety, and 1,3,5-triazine as the tripod splitter module, were designed and synthesized. The potency of 2 was evaluated against MX-1, MCF-7, ID8, L1210FR (BR+, biotin receptor overexpressed) and WI38 (BR-, normal) cell lines in the absence and presence of glutathione (GSH), which is an endogenous thiol that triggers drug release inside the cancer cells. With the GSH and resuspension protocol, 2 exhibited IC50 values of 3.22-9.80 nM against all BR+ cancer cell lines, and 705 nM against WI38. Thus, there was a two orders of magnitude higher selectivity to cancer cells. Also, a clear cooperative effect was observed for the taxoid-camptothecin combination when two drugs were delivered to the cancer cells specifically in the form of a dual-warhead conjugate.
Product Description
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Biotin, meets USP testing specifications
Sigma-Aldrich
Biotin, ≥99.0% (T)
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Biotin, European Pharmacopoeia (EP) Reference Standard
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
BIOTIN
USP
Biotin, United States Pharmacopeia (USP) Reference Standard