Merck
  • Effects of Helicobacter pylori treatment on gastric cancer incidence and mortality in subgroups.
Effects of Helicobacter pylori treatment on gastric cancer incidence and mortality in subgroups.
Journal of the National Cancer Institute (2014-06-14)
Wen-Qing Li, Jun-Ling Ma, Lian Zhang, Linda M Brown, Ji-You Li, Lin Shen, Kai-Feng Pan, Wei-Dong Liu, Yuanreng Hu, Zhong-Xiang Han, Susan Crystal-Mansour, David Pee, William J Blot, Joseph F Fraumeni, Wei-Cheng You, Mitchell H Gail
ABSTRACT
Among 2258 Helicobacter pylori-seropositive subjects randomly assigned to receive one-time H. pylori treatment with amoxicillin-omeprazole or its placebo, we evaluated the 15-year effect of treatment on gastric cancer incidence and mortality in subgroups defined by age, baseline gastric histopathology, and post-treatment infection status. We used conditional logistic and Cox regressions for covariable adjustments in incidence and mortality analyses, respectively. Treatment was associated with a statistically significant decrease in gastric cancer incidence (odds ratio = 0.36; 95% confidence interval [CI] = 0.17 to 0.79) and mortality (hazard ratio = 0.26; 95% CI = 0.09 to 0.79) at ages 55 years and older and a statistically significant decrease in incidence among those with intestinal metaplasia or dysplasia at baseline (odds ratio = 0.56; 95% CI = 0.34 to 0.91). Treatment benefits for incidence and mortality among those with and without post-treatment infection were similar. Thus H. pylori treatment can benefit older members and those with advanced baseline histopathology, and benefits are present even with post-treatment infection, suggesting treatment can benefit an entire population, not just the young or those with mild histopathology.
Product Description
USP
Amoxicillin, United States Pharmacopeia (USP) Reference Standard
Amoxicillin trihydrate, European Pharmacopoeia (EP) Reference Standard
Omeprazole for peak identification, European Pharmacopoeia (EP) Reference Standard
Omeprazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Amoxicillin trihydrate, VETRANAL®, analytical standard
Supelco
Amoxicillin trihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Omeprazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Omeprazole, solid
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis
USP
Omeprazole, United States Pharmacopeia (USP) Reference Standard
Amoxicillin trihydrate for performance verification, European Pharmacopoeia (EP) Reference Standard
Supelco
Omeprazole, analytical standard