Merck
  • Dietary threonine requirement of Pekin ducks from 15 to 35 days of age based on performance, yield, serum natural antibodies, and intestinal mucin secretion.
Dietary threonine requirement of Pekin ducks from 15 to 35 days of age based on performance, yield, serum natural antibodies, and intestinal mucin secretion.
Poultry science (2014-06-07)
Q Zhang, L Xu, A Doster, R Murdoch, P Cotter, A Gardner, T J Applegate
ABSTRACT
A study was conducted to establish the dietary Thr requirement of Pekin ducks from 15 to 35 d of age. Experimental diets were formulated to contain 0.55, 0.60, 0.65, 0.75, and 0.85% Thr (0.57, 0.60, 0.64, 0.72, and 0.80% on an analyzed basis) and were studied in 2 experiments. In experiment 1, each diet was fed to 10 pens of 52 drakes per pen. Samples were collected at d 35 for determinations of carcass yields, serum immune parameters, and intestinal characteristics. Experiment 2 was a digestibility study, wherein 0.5% chromic oxide was mixed into the experimental diets and fed from 15 to 19 d. Ileal digesta were collected at d 19 to analyze mucin secretions and apparent ileal Thr digestibility. The results showed that feeding 0.72% versus 0.64% Thr improved 15 to 35 d BW gain by 55 g (P < 0.05), reduced feed-to-gain by 0.04 (P < 0.05), as well as increased carcass and breast meat yields by 22 and 24 g, respectively. Also, 0.72% Thr had the highest crude mucin secretion on a DM intake (DMI) basis (P < 0.05), although Thr had no effect on villus height, crypt depth, goblet cells, and MUC2 gene expression in the jejunum and ileum. In addition, serum natural IgY linearly increased (P < 0.0001) with dietary Thr increase. Using nonlinear regressions, Thr requirement was estimated to range from a low of 0.70% to maximize dry crude mucin secretion on a DMI basis to a high of 0.80% to maximize carcass weight and serum IgY production by the linear or quadratic regression. Equivalently, Thr requirement varied between a low of 0.62% to minimize mortality and a high of 0.73% to maximize dry crude mucin secretion expressed as DMI using the quadratic broken-line model. Correspondingly, the apparent ileal digestible Thr requirements were estimated to be 0.52 to 0.66% (0.70 to 0.80% dietary Thr) by quadratic and 0.47 to 0.56% (0.62 to 0.73% dietary Thr) by quadratic broken-line model.
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
SAFC
L-Threonine
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
L-Threonine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material