Merck
  • Waterborne cadmium and nickel impact oxidative stress responses and retinoid metabolism in yellow perch.
Waterborne cadmium and nickel impact oxidative stress responses and retinoid metabolism in yellow perch.
Aquatic toxicology (Amsterdam, Netherlands) (2014-06-11)
Michel A Defo, Louis Bernatchez, Peter G C Campbell, Patrice Couture
ABSTRACT
In this experiment, we studied the transcriptional and functional (enzymatic) responses of yellow perch (Perca flavescens) to metal stress, with a focus on oxidative stress and vitamin A metabolism. Juvenile yellow perch were exposed to two environmentally relevant concentrations of waterborne cadmium (Cd) and nickel (Ni) for a period of 6 weeks. Kidney Cd and Ni bioaccumulation significantly increased with increasing metal exposure. The major retinoid metabolites analyzed in liver and muscle decreased with metal exposure except at high Cd exposure where no variation was reported in liver. A decrease in free plasma dehydroretinol was also observed with metal exposure. In the liver of Cd-exposed fish, both epidermal retinol dehydrogenase 2 transcription level and corresponding enzyme activities retinyl ester hydrolase and lecithin dehydroretinyl acyl transferase increased. In contrast, muscle epidermal retinol dehydrogenase 2 transcription level decreased with Cd exposure. Among antioxidant defences, liver transcription levels of catalase, microsomal glutathione-S-transferase-3 and glucose-6-phosphate dehydrogenase were generally enhanced in Cd-exposed fish and this up-regulation was accompanied by an increase in the activities of corresponding enzymes, except for microsomal glutathione-S-transferase. No consistent pattern in antioxidant defence responses was observed between molecular and biochemical response when fish were exposed to Ni, suggesting a non-synchronous response of antioxidant defence in fish exposed to waterborne Ni. There was a general lack of consistency between muscle transcription level and enzyme activities analyzed. The overall findings from this investigation highlight the usefulness of transcriptional and biochemical endpoints in the identification of oxidative stress and vitamin A metabolism impairment biomarkers and the potential use of multi-level biological approaches when assessing environmental risk in fish.
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Nickel, wire, diam. 0.25 mm, ≥99.9%
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2400, slurry, in H2O, active catalyst
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 3202, slurry, in H2O, active catalyst
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 4200, slurry, in H2O, active catalyst
Sigma-Aldrich
Nickel, foil, thickness 0.125 mm, ≥99.9%
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2800, slurry, in H2O, active catalyst
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Nickel, pellets, 20g, max. size 15mm, 99.99+%
Nickel, foil, 10mm disks, thickness 0.038mm, 99.9%
Nickel, foil, 10m coil, thickness 0.125mm, coil width 1mm, as rolled, 99%
Cadmium, rod, 500mm, diameter 5.0mm, as drawn, 99.99+%
Nickel, wire reel, 100m, diameter 0.25mm, annealed, 99%
Nickel, foil, 200x200mm, thickness 0.1mm, annealed, 99%
Nickel, foil, 4mm disks, thickness 0.038mm, 99.9%
Nickel, foil, 4mm disks, thickness 0.05mm, as rolled, 99.98%
Nickel, foil, 2m coil, thickness 0.15mm, coil width 12mm, annealed, 99%
Nickel, wire reel, 0.1m, diameter 1.0mm, as drawn, 99.99+%
Cadmium, foil, not light tested, 25x25mm, thickness 0.001mm, 99.7+%
Cadmium, foil, 8mm disks, thickness 0.5mm, as rolled, 99.95%
Nickel, foil, 10mm disks, thickness 0.025mm, as rolled, 100%
Nickel, foil, 10m coil, thickness 0.3mm, coil width 1mm, as rolled, 99%
Cadmium, tube, 200mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Nickel, foil, 10mm disks, thickness 0.008mm, 99.95%
Nickel, foil, 4mm disks, thickness 0.0075mm, 99.95%
Nickel, wire reel, 1m, diameter 0.75mm, as drawn, 99.99+%
Nickel, foil, 4mm disks, thickness 0.05mm, 99.9%
Nickel, wire reel, 100m, diameter 0.5mm, hard, 99.98%
Nickel, foil, 0.5m coil, thickness 0.25mm, annealed, 99%