Merck
  • Use of a human skin in vitro model to investigate the influence of 'every-day' clothing and skin surface decontamination on the percutaneous penetration of organophosphates.
Use of a human skin in vitro model to investigate the influence of 'every-day' clothing and skin surface decontamination on the percutaneous penetration of organophosphates.
Toxicology letters (2014-06-10)
C A Moore, S C Wilkinson, P G Blain, M Dunn, G A Aust, F M Williams
ABSTRACT
Organophosphates (OPs) are widely used in agriculture. Many studies have investigated the capability of personal protective equipment (PPE) to reduce chemical exposure; however, investigations into the protective effect of 'every-day' clothing are rare. The purpose of this study was to investigate the protective effect of 'every-day' clothing against dermal exposure and to measure early decontamination of skin following exposure to chlorpyrifos and dichlorvos. Using human skin in vitro, absorption of (14)C-labelled chlorpyrifos (500 ng/cm(2)), was shown to be significantly reduced when applied to clothed skin (cotton shirt), regardless of application vehicle (isopropanol (IPA) or propylene glycol (PG)). The majority of applied dose was retained within the clothing after 4 h exposure. Significant reduction in absorption of chlorpyrifos (in PG) was seen through clothed skin when supplemented with skin decontamination at 4 h, compared with clothed skin decontaminated after 24 h, however, this was not observed with IPA. Absorption of dichlorvos (5 μg/cm(2)) was greater through unclothed skin than chlorpyrifos for all vehicles (IPA, isopropyl myristate (IPM) and PG). Significant reduction in absorption was observed when decontaminating clothed skin at 30 min, compared with decontamination at 24 h (post-exposure) for all vehicles. indicate that 'every-day' clothing is effective at reducing exposure to chemicals in contact with skin. Washing the skin surface immediately following removal of exposed clothing can further reduce exposure, depending on the properties of the chemical and vehicle applied.
Product Description
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Propylene glycol, United States Pharmacopeia (USP) Reference Standard
Propylene glycol, European Pharmacopoeia (EP) Reference Standard
Supelco
Chlorpyrifos, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Chlorpyrifos, PESTANAL®, analytical standard
Supelco
Dichlorvos, PESTANAL®, analytical standard
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sigma-Aldrich
(R)-(−)-1,2-Propanediol, 96%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, tested according to Ph. Eur.
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Supelco
1,2-Propanediol, analytical standard
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
Isopropanol, 70% in H2O
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%