Merck
  • Evaluation of a topical gel containing a novel combination of essential oils and antioxidants for reducing oral malodor in dogs.
Evaluation of a topical gel containing a novel combination of essential oils and antioxidants for reducing oral malodor in dogs.
American journal of veterinary research (2014-06-25)
Samuel B Low, R Michael Peak, Christopher W Smithson, Jeanne Perrone, Bert Gaddis, Elias Kontogiorgos
ABSTRACT
To evaluate the effectiveness of a topically applied gel containing essential oils (menthol and thymol) and polyphenolic antioxidants (phloretin and ferulic acid) for reducing halitosis in dogs. 20 dogs. A blinded crossover clinical trial was conducted. Dogs received a dental cleaning and examination (periodontal examination including periodontal probing and assessments of plaque, calculus, and gingivitis). Owners then applied a gel (active or placebo) to oral soft tissues twice daily for a 4-week period. Teeth of the dogs were cleaned again, and owners applied the other gel for a 4-week period. Clinicians scored halitosis immediately after the initial cleaning and at 4 and 8 weeks, and owners scored halitosis weekly. Halitosis assessment by clinicians revealed that both groups had improvement in halitosis scores. Two dogs were removed because of owner noncompliance. In the active-to-placebo group (n = 9), halitosis was significantly reduced during application of the active gel but increased during application of the placebo. Seven of 9 owners reported increased halitosis when treatment was changed from the active gel to the placebo. In the placebo-to-active group (n = 9), halitosis decreased during application of the placebo and continued to decrease during application of the active gel. Seven of 9 owners reported a decrease in halitosis with the active gel. An oral topically applied gel with essential oils and polyphenolic antioxidants applied daily after an initial professional dental cleaning decreased oral malodor in dogs.
Product Description
Menthol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(±)-Menthol, racemic, ≥98.0% (GC)
Sigma-Aldrich
DL-Menthol, ≥95%, FCC, FG
Supelco
DL-Menthol, analytical standard
Thymol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Thymol, FCC, FG
Supelco
Thymol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Menthol, 99%
Sigma-Aldrich
trans-Ferulic acid, 99%
Supelco
Thymol, analytical standard
Supelco
trans-Ferulic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
Thymol, meets analytical specification of Ph. Eur., BP, NF, 99-101%
Sigma-Aldrich
Thymol, ≥98.5%
Sigma-Aldrich
Phloretin, ≥99%
Supelco
4-Hydroxy-3-methoxycinnamic acid, mixture of isomers, analytical standard
Ferulic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Supelco
trans-Ferulic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland