Merck
  • The C-glycosyltransferase IroB from pathogenic Escherichia coli: identification of residues required for efficient catalysis.
The C-glycosyltransferase IroB from pathogenic Escherichia coli: identification of residues required for efficient catalysis.
Biochimica et biophysica acta (2014-06-25)
Daniel Foshag, Cory Campbell, Peter D Pawelek
ABSTRACT
Escherichia coli C-glycosyltransferase IroB catalyzes the formation of a CC bond between enterobactin and the glucose moiety of UDP-glucose, resulting in the production of mono-, di- and tri-glucosylated enterobactin (MGE, DGE, TGE). To identify catalytic residues, we generated a homology model of IroB from aligned structures of two similar C-glycosyltransferases as templates. Superposition of our homology model onto the structure of a TDP-bound orthologue revealed residue W264 as a possible stabilizer of UDP-glucose. D304 in our model was located near the predicted site of the glucose moiety of UDP-glucose. A loop containing possible catalytic residues (H65, H66, E67) was found at the predicted enterobactin-binding site. We generated IroB variants at positions 65-67, 264, and 304 and investigated variant protein conformations and enzymatic activities. Variants were found to have Tm values similar to wild-type IroB. Fluorescence emission spectra of H65A/H66A, E67A, and D304N were superimposable with wild-type IroB. However, the emission spectrum of W264L was blue-shifted, suggesting solvent exposure of W264. While H65A/H66A retained activity (92% conversion of enterobactin, with MGE as a major product), all other IroB variants were impaired in their abilities to glucosylate enterobactin: E67A catalyzed partial (29%) conversion of enterobactin to MGE; W264L converted 55% of enterobactin to MGE; D304N was completely inactive. Activity-impaired variants were found to bind enterobactin with affinities within 2.5-fold of wild-type IroB. Given our outcomes, we propose that IroB W264 and D304 are required for binding and orienting UDP-glucose, while E67, possibly supported by H65/H66, participates in enterobactin/MGE/DGE deprotonation.
Product Description
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tryptophan, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Enterobactin, from Escherichia coli, ≥98% (HPLC)
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
SAFC
L-Aspartic acid
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamic acid hydrochloride, ≥99% (HPLC)
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Aspartic acid, European Pharmacopoeia (EP) Reference Standard
Glutamic acid, European Pharmacopoeia (EP) Reference Standard
Tryptophan, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamic acid, FCC
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Aspartic acid potassium salt, ≥98% (HPLC)
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Phosphorylase b from rabbit muscle, For use as a marker in SDS-PAGE
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
D-Glutamic acid, ≥99% (TLC)
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Phosphorylase b from rabbit muscle, lyophilized powder, ≥20 units/mg protein, 2× crystallization