Merck
  • Exposure to low-dose (56)Fe-ion radiation induces long-term epigenetic alterations in mouse bone marrow hematopoietic progenitor and stem cells.
Exposure to low-dose (56)Fe-ion radiation induces long-term epigenetic alterations in mouse bone marrow hematopoietic progenitor and stem cells.
Radiation research (2014-06-25)
Isabelle R Miousse, Lijian Shao, Jianhui Chang, Wei Feng, Yingying Wang, Antiño R Allen, Jennifer Turner, Blair Stewart, Jacob Raber, Daohong Zhou, Igor Koturbash
ABSTRACT
There is an increasing need to better understand the long-term health effects of high-linear energy transfer (LET) radiation due to exposure during space missions, as well as its increasing use in clinical treatments. Previous studies have indicated that exposure to (56)Fe heavy ions increases the incidence of acute myeloid leukemia (AML) in mice but the underlying molecular mechanisms remain elusive. Epigenetic alterations play a role in radiation-induced genomic instability and the initiation and progression of AML. In this study, we assessed the effects of low-dose (56)Fe-ion irradiation on epigenetic alterations in bone marrow mononuclear cells (BM-MNCs) and hematopoietic progenitor and stem cells (HPSCs). Exposure to (56)Fe ions (600 MeV, 0.1, 0.2 and 0.4 Gy) resulted in significant epigenetic alterations involving methylation of DNA, the DNA methylation machinery and expression of repetitive elements. Four weeks after irradiation, these changes were primarily confined to HPSCs and were exhibited as dose-dependent hypermethylation of LINE1 and SINE B1 repetitive elements [4.2-fold increase in LINE1 (P < 0.001) and 7.6-fold increase in SINE B1 (P < 0.01) after exposure to 0.4 Gy; n = 5]. Epigenetic alterations were persistent and detectable for at least 22 weeks after exposure, when significant loss of global DNA hypomethylation (1.9-fold, P < 0.05), decreased expression of Dnmt1 (1.9-fold, P < 0.01), and increased expression of LINE1 and SINE B1 repetitive elements (2.8-fold, P < 0.001 for LINE1 and 1.9-fold, P < 0.05 for SINE B1; n = 5) were observed after exposure to 0.4 Gy. In contrast, exposure to (56)Fe ions did not result in accumulation of increased production of reactive oxygen species (ROS) and DNA damage, exhibited as DNA strand breaks. Furthermore, no significant alterations in cellular senescence and apoptosis were detected in HPSCs after exposure to (56)Fe-ion radiation. These findings suggest that epigenetic reprogramming is possibly involved in the development of radiation-induced genomic instability and thus, may have a causative role in the development of AML.
Sigma-Aldrich
Carbonyl iron, ≥97% Fe basis
Iron, rod, 1000mm, diameter 6.35mm, as drawn, soft ingot 99.8%
Sigma-Aldrich
Iron, foil, thickness 0.25 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Iron, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, granular, 10-40 mesh, >99.99% trace metals basis
Iron, rod, 200mm, diameter 80mm, as drawn, armcO« soft ingot 99.8%
Iron, foil, not light tested, 50x50mm, thickness 0.004mm, 99.85%
Iron, foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%
Iron, foil, 2m coil, thickness 0.5mm, hard, 99.5%
Iron, foil, 150x150mm, thickness 5mm, as rolled, 99.8+%
Sigma-Aldrich
Iron, ≥99%, reduced, powder (fine)
Iron, foil, not light tested, 25x25mm, thickness 0.005mm, 99.85%
Iron, foil, 150x150mm, thickness 3.0mm, as rolled, 99.5%
Iron, foil, 0.5m coil, thickness 0.05mm, hard, 99.5%
Iron, foil, 10m coil, thickness 0.15mm, hard, 99.5%
Iron, foil, 50x50mm, thickness 1.83mm, Quarter hard, 99.5%
Iron, tube, 100mm, outside diameter 5.0mm, inside diameter 4.5mm, wall thickness 0.25mm, as drawn, 99.5%
Iron, wire reel, 2m, diameter 0.13mm, hard, 99.99+%
Iron, wire reel, 10m, diameter 0.065mm, hard, 99.5%
Iron, tube, 500mm, outside diameter 4.0mm, inside diameter 3.6mm, wall thickness 0.2mm, as drawn, 99.5%
Iron, wire reel, 5m, diameter 0.13mm, hard, 99.99+%
Iron, tube, 100mm, outside diameter 2.0mm, inside diameter 1.08mm, wall thickness 0.46mm, as drawn, 99.5%
Iron, foil, 25x25mm, thickness 2.0mm, as rolled, 99.95%
Iron, wire reel, 0.5m, diameter 0.5mm, hard, 99.99+%
Iron, tube, 1000mm, outside diameter 4.0mm, inside diameter 3.6mm, wall thickness 0.2mm, as drawn, 99.5%
Iron, tube, 500mm, outside diameter 20.0mm, inside diameter 16mm, wall thickness 2.0mm, annealed, 99.5%
Iron, foil, 10mm disks, thickness 0.004mm, 99.85%
Iron, tube, 200mm, outside diameter 3.2mm, inside diameter 2.2mm, wall thickness 0.5mm, as drawn, 99.8%
Iron, foil, 0.2m coil, thickness 0.5mm, coil width 49mm, armco« soft ingot 99.8+%
Iron, wire reel, 10m, diameter 0.5mm, as drawn, 99.95%