Merck
Morphine glucuronidation increases its analgesic effect in guinea pigs.
Life sciences (2014-06-27)
Ana Oliveira, Dora Pinho, António Albino-Teixeira, Rui Medeiros, Ricardo Jorge Dinis-Oliveira, Félix Carvalho
ABSTRACT
Morphine is extensively metabolized to neurotoxic morphine-3-glucuronide (M3G) and opioid agonist morphine-6-glucuronide (M6G). Due to these different roles, interindividual variability and co-administration of drugs that interfere with metabolism may affect analgesia. The aim of the study was to investigate the repercussions of administration of an inducer (2,3,7,8-tetrachlorodibenzo-p-dioxin, TCDD) and an inhibitor (ranitidine) of glucuronidation in morphine metabolism and consequent analgesia, using the Guinea pig as a suitable model. Thirty male Dunkin-Hartley guinea pigs were divided in six groups: control, morphine, ranitidine, ranitidine+morphine, TCDD and TCDD+morphine. After previous exposure to TCDD and ranitidine, morphine effect was assessed by an increasing temperature hotplate (35-52.5°C), during 60min after morphine administration. Then, blood was collected and plasma morphine and metabolites were quantified. Animals treated with TCDD presented faster analgesic effect and 75% reached the cut-off temperature of 52.5°C, comparing with only 25% in morphine group. Animals treated with ranitidine presented a significantly lower analgesic effect, compared with morphine group (p<0.05). Moreover, significant differences between groups were found in M3G levels and M3G/morphine ratio (p<0.001 and p<0.0001), with TCDD animals presenting the highest values for M3G, M6G, M3G/morphine and M6G/morphine, and the lowest value for morphine. The opposite was observed in the animals treated with ranitidine. Our results indicate that modulation of morphine metabolism may result in variations in metabolite concentrations, leading to different analgesic responses to morphine, in an animal model that may be used to improve morphine effect in clinical practice.
Sigma-Aldrich
Phenacetin, ≥98.0% (HPLC)
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
USP
Phenacetin Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
Supelco
Phenacetin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%