Merck
  • Early hysteroscopic removal of intratubal microinserts with urologic stone retrieval forceps.
Early hysteroscopic removal of intratubal microinserts with urologic stone retrieval forceps.
Obstetrics and gynecology (2014-07-09)
Lisa M Goldthwaite, Lindsay Edwards, Kristina Tocce
ABSTRACT
Nickel hypersensitivity reactions can be an indication for the removal of intratubal microinserts used for hysteroscopic sterilization. Although early removal can be attempted hysteroscopically, hysteroscopic grasping forceps may be inadequate to grasp deeply positioned inserts. Three days after hysteroscopic sterilization, the patient presented with a rash consistent with a nickel hypersensitivity reaction. Ten days after placement, we successfully performed hysteroscopic removal of an intratubal microinsert with tri-prong urologic stone retrieval forceps after standard hysteroscopic grasping forceps was unable to reach the deeply positioned device. Within 36 hours of bilateral microinsert removal, all symptoms resolved. In difficult cases, 1-mm tri-prong urologic stone retrieval forceps can be useful for early hysteroscopic removal of intratubal microinserts.
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Nickel, pellets, 50g, max. size 10mm, 99.999%
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Nickel, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99+%
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Nickel, foil, light tested, 100x100mm, thickness 0.025mm, 99.9%
Nickel, wire reel, 5m, diameter 0.025mm, as drawn, 99.98%
Nickel, foil, 5m coil, thickness 0.05mm, annealed, 99%
Nickel, wire reel, 0.1m, diameter 0.75mm, as drawn, 99.99+%
Nickel, foil, 0.5m coil, thickness 0.025mm, 99.9%
Nickel, rod, 500mm, diameter 2.0mm, 99.99+%
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Nickel, foil, 0.5m coil, thickness 0.0075mm, 99.95%
Nickel, foil, 0.2m coil, thickness 0.005mm, as rolled, 99.9%
Nickel, rod, 200mm, diameter 10.0mm, 99.99+%
Nickel, foil, light tested, 100x100mm, thickness 0.05mm, 99.9%
Nickel, foil, 0.5m coil, thickness 0.0125mm, 99.9%
Nickel, foil, not light tested, 300x300mm, thickness 0.0125mm, 99.9%
Nickel, foil, not light tested, 25x25mm, thickness 0.001mm, 99.95%
Nickel, foil, 1m coil, thickness 0.075mm, annealed, 99%
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Nickel, mesh, 30x30mm, nominal aperture 0.34mm, thickness 0.025mm, wire diameter 0.041mm, 70 wires/inch, open area 80%, electro-formed mesh, 99.9%
Nickel, rod, 1000mm, diameter 3.0mm, 99%
Nickel, foil, 0.5m coil, thickness 0.125mm, annealed, 99%
Nickel, foil, 10mm disks, thickness 0.05mm, 99.9%
Nickel, foil, light tested, 300x300mm, thickness 0.02mm, 99.9%
Nickel, foil, light tested, 300x300mm, thickness 0.025mm, 99.9%
Nickel, wire reel, 5m, diameter 2.0mm, hard, 99.98%
Nickel, foil, light tested, 100x100mm, thickness 0.007mm, 99.95%