Merck
  • The use of a dermal substitute for simultaneous flap delay and donor site coverage in two cases.
The use of a dermal substitute for simultaneous flap delay and donor site coverage in two cases.
Journal of wound care (2014-08-27)
Brittany J Behar, Hamid Abdollahi, Bharat Ranganath, Azra Ashraf, Paul M Glat
ABSTRACT
Lower extremity traumatic wounds can be difficult to treat owing to limb ischaemia and large zones of injury. Often, muscle or fasciocutaneous flaps are used in the presence of severe open orthopaedic injuries with soft tissue defects. Sometimes local flaps may be the preferred or only option, but may not tolerate being rotated or advanced owing to resulting flap ischaemia. One well-studied technique that can increase the survival of various flaps involves the delay phenomenon. In these case reports, Integra Dermal Regeneration Template was used to simultaneously create delayed flaps and to cover the wound and flap donor site so that the donor site could be skin grafted at the time of flap inset. These cases demonstrate that use of Integra can enhance the delay phenomenon while simultaneously providing coverage of soft tissue defects in preparation for ultimately insetting delayed flaps and better covering donor areas. This technique may be applicable to many different flaps in many different anatomic locations and should be considered an option when reconstructing complicated wounds.
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Chondroitin 4-sulfate sodium salt from bovine trachea, mixture of isomers, main component: chondroitin 4-sulfate
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Chondroitin sulfate A sodium salt from bovine trachea, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance