Merck
  • Photodegradation study of nystatin by UV-Vis spectrophotometry and chemometrics modeling.
Photodegradation study of nystatin by UV-Vis spectrophotometry and chemometrics modeling.
Journal of AOAC International (2014-08-26)
Bahram Hemmateenejad, Mohsen Nekoeinia, Ghodratollah Absalan, Mehdi Ansari
ABSTRACT
Nystatin, one of the tetraene antifungal antibiotics, is very sensitive to light. Thus, when nystatin is exposed to natural daylight, it is photodegraded to products of lower biological activity. In this work, the photodegradation kinetics of nystatin was monitored by a UV-Vis spectrophotometry method. The absorbance spectra of the nystatin, exposed to a 366 nm UV lamp, were recorded at different periods of time. By application of factor analysis to the absorbance data matrix, three absorbing chemical species, coexisting in the reaction system, were detected. The soft-modeling multivariate curve resolution-alternating least squares analysis of the evolutionary absorbance data revealed that nystatin undergoes photodegradation in a two-step consecutive manner. Hard-modeling data analysis suggested that reaction has first-order kinetics in the first step and zero-order kinetics in the second step. The reaction rate constants of the first and second steps were estimated as 0.0929 (+/-0.0076) and 0.0052 (+/-0.0016)/min, respectively. Finally, the pure spectra of the resolved chemical species were calculated.
Sigma-Aldrich
Nystatin Suspension, suspension, 10,000 unit/mL in DPBS, BioReagent, suitable for cell culture
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Nystatin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Nystatin, powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Nystatin, ≥4,400 units/mg
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Nystatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
USP
Nystatin, United States Pharmacopeia (USP) Reference Standard