Merck
  • Formation kinetics of gemfibrozil chlorination reaction products: analysis and application.
Formation kinetics of gemfibrozil chlorination reaction products: analysis and application.
Water environment research : a research publication of the Water Environment Federation (2014-08-13)
Wendy H Krkosek, Sigrid Peldszus, Peter M Huck, Graham A Gagnon
ABSTRACT
Aqueous chlorination kinetics of the lipid regulator gemfibrozil and the formation of reaction products were investigated in deionized water over the pH range 3 to 9, and in two wastewater matrices. Chlorine oxidation of gemfibrozil was found to be highly dependent on pH. No statistically significant degradation of gemfibrozil was observed at pH values greater than 7. Gemfibrozil oxidation between pH 4 and 7 was best represented by first order kinetics. At pH 3, formation of three reaction products was observed. 4'-C1Gem was the only reaction product formed from pH 4-7 and was modeled with zero order kinetics. Chlorine oxidation of gemfibrozil in two wastewater matrices followed second order kinetics. 4'-C1Gem was only formed in wastewater with pH below 7. Deionized water rate kinetic models were applied to two wastewater effluents with gemfibrozil concentrations reported in literature in order to calculate potential mass loading rates of 4'C1Gem to the receiving water.
Sigma-Aldrich
Chlorine, puriss., ≥99.8%
Sigma-Aldrich
Chlorine, ≥99.5%
Supelco
Gemfibrozil, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Gemfibrozil
Gemfibrozil for system suitability, European Pharmacopoeia (EP) Reference Standard
Gemfibrozil, European Pharmacopoeia (EP) Reference Standard