Merck
  • Determining the minimum inhibitory concentration of Tetraclean against Candida albicans.
Determining the minimum inhibitory concentration of Tetraclean against Candida albicans.
Nigerian journal of medicine : journal of the National Association of Resident Doctors of Nigeria (2014-09-05)
A Qyasian, Z Mohammadi, L Giardino, F Palazzi, S Shalavi, S Sabbaghi, E Khoshbin
ABSTRACT
The purpose of the present study was to determine the minimum inhibitory concentrations of Tetraclean, chlorhexidine, hydrogen peroxide, and sodium hypochlorite against Candida albicans.Amphotericin B was used as positive control and RPMI plus 1 ml Candida suspension was used as negative control. Serial dilution method was used to determine MIC of the irrigants. Findings showed that all positive controls demonstrated complete inhibition of C. albicans at concentration of 0.78 microg mL(-1). On the other hand, all negative controls were positive for fungal growth which confirms the methodology of the study. Findings showed that the MIC of CHX was significantly lesser than other tested irrigants (p < 0.05). The MICs of other groups in an ascending order were as follows: Tetraclean, NaOCl, and H2O2. However, the difference betweenTetraclean and NaOCI was not significant (p > 0.05). It can be concluded that MIC of CHX was significantly lower than other irrigations solutions which confirms its strong antifungal activity.
Product Description
Supelco
Doxycycline hyclate, Pharmaceutical Secondary Standard; Certified Reference Material
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Doxycycline hyclate
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Supelco
Doxycycline hyclate, VETRANAL®, analytical standard
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
USP
Doxycycline hyclate, United States Pharmacopeia (USP) Reference Standard
Doxycycline hyclate, European Pharmacopoeia (EP) Reference Standard
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Chlorhexidine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Chlorhexidine, ≥99.5%
Sigma-Aldrich
Chlorhexidine dihydrochloride, ≥98%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Doxycycline monohydrate
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, 99%
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
Chlorhexidine, European Pharmacopoeia (EP) Reference Standard