Merck
  • Expression of SLP-2 was associated with invasion of esophageal squamous cell carcinoma.
Expression of SLP-2 was associated with invasion of esophageal squamous cell carcinoma.
PloS one (2013-05-15)
Wenfeng Cao, Bin Zhang, Fang Ding, Weiran Zhang, Baocun Sun, Zhihua Liu
ABSTRACT
Stomatin-like protein 2 (SLP-2), a member of the Stomatin superfamily, has been identified as an oncogenic-related protein and found to be up-regulated in multi-cancers. Nonetheless, the expression pattern and regulation of SLP-2 in human esophageal squamous cell carcinoma (ESCC) remain unexplored. Immunohistochemistry and immunofluorescence staining analysis were performed to show SLP-2 expression and location. RNAi method was used to inhibit specific protein expression. Transwell assay was done to investigate cells invasive capability. RT-PCR and Western blot analysis were used to detect mRNA and protein expression levels. Immunohistochemical analysis showed that up-regulation of SLP-2 was found in invasive front compared with cancer central tissue in ESCC. Inhibition of SLP-2 by SLP-2 siRNA can decrease ESCC cells invasive capability through MMP-2 dependent manner. Up-regulation of SLP-2 was effectively abrogated by the ERK1/2 inhibitors either PD98059 or U0126, but no effect was showed by the treatment of AKT inhibitors either LY294002 or MK-2206. So the regulation of SLP-2 was involved in activation of the MAPK/ERK pathway. We found that PMA/EGF could induce the up-regulated expression of SLP-2 probably through activating ERK signalling. The current study suggests that SLP-2 may represent an important molecular hallmark that is clinically relevant to the invasion of ESCC.
Sigma-Aldrich
MISSION® esiRNA, targeting human STOML2