Merck
  • Characterization of the low-temperature activity of Sulfolobus tokodaii glucose-1-dehydrogenase mutants.
Characterization of the low-temperature activity of Sulfolobus tokodaii glucose-1-dehydrogenase mutants.
Journal of bioscience and bioengineering (2014-04-20)
Taisuke Sugii, Satoshi Akanuma, Sota Yagi, Kazuki Yagyu, Yukiko Shimoda, Akihiko Yamagishi
ABSTRACT
Thermophilic enzymes are potentially useful for industrial processes because they are generally more stable than are mesophilic or psychrophilic enzymes. However, a crucial drawback for their use in such processes is that most thermophilic enzymes are nearly inactive at moderate and low temperatures. We have previously proposed that modulation of the coenzyme-binding pocket of thermophilic dehydrogenases can produce mutated proteins with enhanced low-temperature activities. In the current study, we produced and characterized mutants of an NADP-dependent glucose-1-dehydrogenase from the hyperthermophile Sulfolobus tokodaii in which a predicted coenzyme-binding, non-polar residue was replaced by another non-polar residue. Detailed analyses of the kinetic properties of the wild-type enzyme and its mutants showed that one of the mutants (V254I) had improved kcat and kcat/Km values at both 25°C and 80°C. Temperature-induced unfolding experiments showed that the thermal stability of the mutant enzyme was comparable to that of the wild-type enzyme. Calculation of the energetic contribution of the V254I mutation for the dehydrogenase reaction revealed that the mutation destabilizes the enzyme-NADP(+)-glucose ternary complex and reduces the transition-state energy, thus enhancing catalysis.
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
Sigma-Aldrich
Glucose dehydrogenase from Pseudomonas sp., powder, white, ≥200 U/mg
Sigma-Aldrich
L-Isoleucine, BioUltra, ≥99.5% (NT)
SAFC
L-Isoleucine
Sigma-Aldrich
L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Isoleucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate hydrate
Supelco
L-Isoleucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
SAFC
L-Valine
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Valine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
Isoleucine, European Pharmacopoeia (EP) Reference Standard