Merck
  • Micropatterned culture of HepG2 spheroids using microwell chip with honeycomb-patterned polymer film.
Micropatterned culture of HepG2 spheroids using microwell chip with honeycomb-patterned polymer film.
Journal of bioscience and bioengineering (2014-04-20)
Hidekazu Yamazaki, Shun Gotou, Koju Ito, Souichi Kohashi, Yuki Goto, Yukiko Yoshiura, Yusuke Sakai, Hiroshi Yabu, Masatsugu Shimomura, Kohji Nakazawa
ABSTRACT
Microwell chip culture is a promising technique for the generation of homogenous spheroids. We investigated the relationship between the structure of the bottom surface of microwell chip and the properties of HepG2 spheroid. We developed a microwell chip, the bottom surface of which consisted of a honeycomb-patterned polymer film (honeycomb film) that had a regular porous structure (HF chip). The chip comprised 270 circular microwells; each microwell was 600 μm in diameter and 600 μm in depth. At the center of the honeycomb film, an area, 200 μm in diameter, was modified with collagen to facilitate cell adhesion. With the exception of the collagen-coated area, the entire microwell was modified with polyethylene glycol to eliminate cell adhesion. HepG2 cells formed uniform spheroids when cultured in the microwells of HF chip. Furthermore, the cells passed through the porous structure of honeycomb film and formed spheroids at its opposite side. The spheroid growth of HepG2 cells cultured in HF chip was greater than that when the cells were culture in a microwell chip, the bottom surface of which was made of poly-methylmethacrylate (PMMA chip). The albumin secretion activity of HepG2 spheroids in HF chip was equal to that in PMMA chip. These results indicate that the microwell bottom with a porous structure enhances the cell growth and maintains well the spheroid function. Thus, HF chip is a promising platform for spheroid cell culture.
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Poly(methyl methacrylate), isotactic, >80% isotactic
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~350,000 by GPC
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 20,000
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~15,000 by GPC, powder
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), average Mw 97,000 (Typical), average Mn 46,000 (Typical)
Supelco
Poly(methyl methacrylate), analytical standard, 2,480,000
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 100,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 2,000
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 8,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 10,000