Merck
  • Differential regeneration of myocardial infarction depending on the progression of disease and the composition of biomimetic hydrogel.
Differential regeneration of myocardial infarction depending on the progression of disease and the composition of biomimetic hydrogel.
Journal of bioscience and bioengineering (2014-05-27)
So Jeong Yoon, Soyoung Hong, Yong Hu Fang, Myeongjin Song, Kuk Hui Son, Ho Sung Son, Sook Kyoung Kim, Kyung Sun, Yongdoo Park
ABSTRACT
Hydrogel has been used for regenerating myocardial infraction (MI) as a delivery vehicle for cells and growth factors. This study showed that injectable hyaluronic acid (HA)-based hydrogels alone would effectively regenerate the damaged infarcted heart tissue. We found that there are two major factors of regeneration in MI. One is molecular weight of HA and another is the progression of MI; sub-acute and chronic. Rat MI model was prepared by ligating the left anterior descending coronary artery (LAD). Four weeks after injection of hydrogel, functional analysis of the heart and histological analysis was assessed. When different molecular weight HA-based hydrogels with 50 kDa, 130 kDa, and 170 kDa were applied to the infarcted area in the sub-acute model, 50 kDa HA-based hydrogel showed the most significant regeneration of myocardium as well as functional recovery among samples. For the disease progression, 50 kDa HA-based hydrogels were injected to sub-acute and chronic MI models. The regeneration activity was significantly decreased in the chronic models reflecting that injection timing of the therapeutic agents is also major determinants in the regeneration process. These results suggest that injection time and composition of hydrogel are two major points treating MI.
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 15,000-30,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,400,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,000,000-1,250,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 300,000-500,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 50,000-90,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 8,000-15,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,500,000-1,750,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 10,000-30,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 30,000-50,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,200
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,200,000
Sigma-Aldrich
Acrylic acid N-hydroxysuccinimide ester, ≥90% (HPLC)
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus zooepidemicus, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from rooster comb, avian glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 90,000-130,000
Supelco
Triethanolamine, analytical standard
Sigma-Aldrich
Triethanolamine, puriss. p.a., ≥99% (GC)
Sigma-Aldrich
Adipic acid dihydrazide, ≥98% (titration)
Sigma-Aldrich
Triethanolamine, puriss., meets analytical specification of NF, ≥99% (GC)
Sigma-Aldrich
Triethanolamine, reagent grade, 98%
Sigma-Aldrich
Triethanolamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethanolamine, ≥99.0% (GC)
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 130,000-150,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 750,000-1,000,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 500,000-750,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,250,000-1,500,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 150,000-300,000
Sigma-Aldrich
1-Hydroxybenzotriazole hydrate, wetted with not less than 20 wt. % water, 97% dry basis
Sigma-Aldrich
1-Hydroxybenzotriazole hydrate, wetted with not less than 14 wt. % water, 98% dry basis