Merck
Odour emission characteristics of 22 recreational rivers in Nanjing.
Environmental monitoring and assessment (2014-06-19)
Yu Wan, Xiaohong Ruan, Xinguang Wang, Qian Ma, Xiaoming Lu
ABSTRACT
The odour emission characteristics of 22 recreational rivers in Nanjing were investigated and analysed. Eight odorous compounds (ammonia (NH₃), hydrogen sulphide (H₂S), sulphur dioxide (SO₂), carbon disulphide (CS₂), nitrobenzene (C₆H₅NO₂), aniline (C₆H₅NH₂), dimethylamine (C₂H₇N), and formaldehyde (HCHO)) were measured in odour emission samples collected using a custom-made emission flux hood chamber. The results showed that all odorants were detected in all monitoring rivers. NH₃ was the main odorant, with emission rates ranging from 4.86 to 15.13 μg/min m(2). The total odour emission rate of the Nan River, at 1 427.07 OU/s, was the highest of the all investigated rivers. H₂S, NH₃ and nitrobenzene were three key odour emission contributors according to their contributions to the total odour emission. A correlation analysis of the pollutants showed there was a significant positive correlation between the emission rate of NH₃ and the concentration of ammonia nitrogen (NH₄ (+)-N) and total nitrogen (TN). The H₂S and SO₂ emission rates had a significant positive correlation with sulphides (S(2-)) and available sulphur (AS) in the water and sediment. The content of TN, NH₄(+)-N, S(2-) and AS in the water and sediment affected the concentration of H₂S, SO₂ and NH₃ in the emission gases. NH₄(+)-N, S(2-) and AS are suggested as the key odour control indexes for reducing odours emitted from these recreational rivers. The study provides useful information for effective pollution control, especially for odour emission control for the recreational rivers of the city. It also provides a demonstrate example to show how to monitor and assess a contaminated river when odour emission and its control need to be focused on.
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Ammonia solution, 7 N in methanol