Merck
  • Hypoxia inducible factor (HIF)-1α directly activates leptin receptor (Ob-R) in pancreatic cancer cells.
Hypoxia inducible factor (HIF)-1α directly activates leptin receptor (Ob-R) in pancreatic cancer cells.
Cancer letters (2014-08-19)
He Ren, Lingling Jia, Tiansuo Zhao, Huan Zhang, Jing Chen, Shaoguang Yang, Jingcheng Liu, Ming Yu, Jihui Hao
ABSTRACT
The aim of this study is to investigate the regulatory mechanism of leptin receptors (Ob-R) in pancreatic cancer. We found that the over-expression of hypoxia inducible factor (HIF-1)α and hypoxia up-regulated the expression of Ob-R in pancreatic cancer cells. When HIF-1α gene was silenced in vitro, the expression of Ob-R was significantly decreased. Xenograft mouse models showed that the inhibition of HIF-1α resulted in the concomitant decrease of Ob-R in vivo. In addition, HIF-1α expression was correlated with Ob-R in pancreatic cancer tissues by immunohistochemical staining. Clinical data showed that over-expression of HIF-1 was associated with pathological tumor node metastasis stage, lymph node metastasis and overall survival. HIF-1α directly bound to the hypoxia-responsive element (HRE) located in Ob-R gene promoter (-828/-832) and activated the transcription. Finally, we demonstrated that the silence of HIF-1α gene reversed the inhibitory effect of leptin/Ob-R in pancreatic cancer cells. Taken together, our results indicate that HIF-1α directly regulated Ob-R expression in pancreatic cancer, which might be a valuable therapeutic target for pancreatic cancer.
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance