Merck
  • Brentuximab vedotin in the front-line treatment of patients with CD30+ peripheral T-cell lymphomas: results of a phase I study.
Brentuximab vedotin in the front-line treatment of patients with CD30+ peripheral T-cell lymphomas: results of a phase I study.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2014-08-20)
Michelle A Fanale, Steven M Horwitz, Andres Forero-Torres, Nancy L Bartlett, Ranjana H Advani, Barbara Pro, Robert W Chen, Andrew Davies, Tim Illidge, Dirk Huebner, Dana A Kennedy, Andrei R Shustov
ABSTRACT
Front-line treatment of peripheral T-cell lymphomas (PTCL) involves regimens such as cyclophosphamide, doxorubicin, vincristine, prednisone (CHOP) and results in a 5-year overall survival (OS) rate of less than 50%. This phase I open-label study evaluated the safety and activity of brentuximab vedotin administered sequentially with CHOP or in combination with CHP (CHOP without vincristine) as front-line treatment in patients with CD30(+) PTCL. Patients received sequential treatment (once every 3 weeks) with brentuximab vedotin 1.8 mg/kg (two cycles) followed by CHOP (six cycles) or brentuximab vedotin 1.8 mg/kg plus CHP (BV+CHP) for six cycles (once every 3 weeks). Responders received single-agent brentuximab vedotin for eight to 10 additional cycles (for a total of 16 cycles). The primary objective was assessment of safety; secondary end points included objective response rate, complete remission (CR) rate, progression-free survival rate (PFS), and OS. There were no prespecified comparisons of the two treatment approaches. After sequential treatment, 11 (85%) of 13 patients achieved an objective response (CR rate, 62%; estimated 1-year PFS rate, 77%). Grade 3/4 adverse events occurred in eight (62%) of 13 patients. At the end of combination treatment, all patients (n = 26) achieved an objective response (CR rate, 88%; estimated 1-year PFS rate, 71%). All seven patients without anaplastic large-cell lymphoma achieved CR. Grade 3/4 adverse events (≥ 10%) in the combination-treatment group were febrile neutropenia (31%), neutropenia (23%), anemia (15%), and pulmonary embolism (12%). Brentuximab vedotin, administered sequentially with CHOP or in combination with CHP, had a manageable safety profile and exhibited substantial antitumor activity in newly diagnosed patients with CD30(+) PTCL. A randomized phase III trial is under way, comparing BV+CHP with CHOP (clinical trial No. NCT01777152).
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Vincristine sulfate, meets USP testing specifications
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Prednisone, ≥98%
Supelco
Prednisone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Doxorubicin hydrochloride, United States Pharmacopeia (USP) Reference Standard
Vincristine sulfate, European Pharmacopoeia (EP) Reference Standard
Doxorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Prednisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Vincristine sulfate salt, 95.0-105.0% (HPLC), powder or crystals