Merck
  • Large sulfur isotope fractionations associated with Neoarchean microbial sulfate reduction.
Large sulfur isotope fractionations associated with Neoarchean microbial sulfate reduction.
Science (New York, N.Y.) (2014-11-08)
Iadviga Zhelezinskaia, Alan J Kaufman, James Farquhar, John Cliff
ABSTRACT
The minor extent of sulfur isotope fractionation preserved in many Neoarchean sedimentary successions suggests that sulfate-reducing microorganisms played an insignificant role in ancient marine environments, despite evidence that these organisms evolved much earlier. We present bulk, microdrilled, and ion probe sulfur isotope data from carbonate-associated pyrite in the ~2.5-billion-year-old Batatal Formation of Brazil, revealing large mass-dependent fractionations (approaching 50 per mil) associated with microbial sulfate reduction, as well as consistently negative Δ(33)S values (~ -2 per mil) indicative of atmospheric photochemical reactions. Persistent (33)S depletion through ~60 meters of shallow marine carbonate implies long-term stability of seawater sulfate abundance and isotope composition. In contrast, a negative Δ(33)S excursion in lower Batatal strata indicates a response time of ~40,000 to 150,000 years, suggesting Neoarchean sulfate concentrations between ~1 and 10 μM.
Product Description
Iron, rod, 50mm, diameter 6.0mm, as drawn, 99.99+%
Iron, foil, 10mm disks, thickness 0.0125mm, 99.99+%
Iron, foil, 100x100mm, thickness 0.5mm, as rolled, 99.99+%
Iron, tube, 1000mm, outside diameter 6.35mm, inside diameter 5.35mm, wall thickness 0.5mm, as drawn, 99.8%
Iron, foil, 50x50mm, thickness 1.83mm, Quarter hard, 99.5%
Iron, wire reel, 2m, diameter 0.13mm, hard, 99.99+%
Iron, tube, 1000mm, outside diameter 12.7mm, inside diameter 9.5mm, wall thickness 1.6mm, annealed, 99.5%
Iron, tube, 100mm, outside diameter 1.65mm, inside diameter 0.89mm, wall thickness 0.38mm, as drawn, 99.8+%
Iron, rod, 50mm, diameter 8.0mm, as drawn, 99.99+%
Iron, foil, 10mm disks, thickness 0.015mm, 99.85%
Iron, rod, 500mm, diameter 3.2mm, as drawn, 98+%
Iron, tube, 500mm, outside diameter 10.0mm, inside diameter 7mm, wall thickness 1.5mm, annealed, 99.5%
Iron, foil, 25x25mm, thickness 3.1mm, as rolled, 99.95%
Iron, tube, 100mm, outside diameter 2.0mm, inside diameter 1.08mm, wall thickness 0.46mm, as drawn, 99.5%
Iron, rod, 200mm, diameter 40mm, as drawn, armcO« soft ingot 99.8%
Iron, rod, 500mm, diameter 4.8mm, as drawn, 98+%
Iron, tube, 200mm, outside diameter 2.30mm, inside diameter 2.07mm, wall thickness 0.115mm, as drawn, 99.5%
Iron, tube, 200mm, outside diameter 2.0mm, inside diameter 1.08mm, wall thickness 0.46mm, as drawn, 99.5%
Iron, tube, 1000mm, outside diameter 4.0mm, inside diameter 3.6mm, wall thickness 0.2mm, as drawn, 99.5%
Iron, foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%
Iron, foil, 100x100mm, thickness 0.5mm, hard, 99.5%
Iron, foil, 100x100mm, thickness 0.38mm, hard, 99.5%
Iron, tube, 500mm, outside diameter 20.0mm, inside diameter 16mm, wall thickness 2.0mm, annealed, 99.5%
Iron, rod, 500mm, diameter 12.7mm, as drawn, 98+%
Iron, foil, 50x50mm, thickness 2.0mm, as rolled, 99.95%
Iron, foil, 10mm disks, thickness 0.006mm, 99.85%
Iron, foil, 100x100mm, thickness 1.83mm, Quarter hard, 99.5%
Iron, rod, 500mm, diameter 2.0mm, as drawn, 99.99+%
Iron, rod, 200mm, diameter 30mm, as drawn, armcO« soft ingot 99.8%
Iron, tube, 200mm, outside diameter 1.65mm, inside diameter 0.89mm, wall thickness 0.38mm, as drawn, 99.8+%