Merck
  • Transepithelial iontophoresis corneal collagen cross-linking for progressive keratoconus: initial clinical outcomes.
Transepithelial iontophoresis corneal collagen cross-linking for progressive keratoconus: initial clinical outcomes.
Journal of refractive surgery (Thorofare, N.J. : 1995) (2014-11-07)
Paolo Vinciguerra, J Bradley Randleman, Vito Romano, Emanuela F Legrottaglie, Pietro Rosetta, Fabrizio I Camesasca, Raffaele Piscopo, Claudio Azzolini, Riccardo Vinciguerra
ABSTRACT
To report initial clinical results of transepithelial corneal collagen cross-linking with iontophoresis (I-CXL). Twenty eyes of 20 patients diagnosed as having progressive keratoconus who underwent I-CXL were included in this prospective non-randomized clinical study. Corrected distance visual acuity (CDVA), spherical equivalent and cylinder refraction, various corneal topography and Scheimpflug tomography parameters, aberrometry, anterior segment optical coherence tomography, and endothelial cell count were assessed at baseline and at 1, 3, 6, and 12 months postoperatively. CDVA improved significantly at 3, 6, and 12 months postoperatively (logMAR difference of -0.07 ± 0.01, -0.09 ± 0.03, and -0.12 ± 0.06, respectively; P < .05). Aberrometry remained stable during follow-up and a trend toward improvement was noted. All topographic parameters (including maximum keratometry) were stable during the follow-up, but exhibited a positive non-significant trend toward improvement. Minimum corneal thickness values were stable for up to 12 months postoperatively. None of the patients showed a progression of keratoconus. Endothelial cell counts did not change significantly (P > .05). Preliminary results up to 1 year postoperatively indicate the efficacy of I-CXL in stabilizing the progression of this degenerative disease combined with significant improvement of CDVA. I-CXL, which spares the corneal epithelium, has the potential to become a valid alternative for halting the progression of keratoconus while reducing postoperative patient pain, risk of infection, and treatment time in select patients; however, the relative efficacy of this technique compared to standard epithelium-off techniques remains to be determined.
Product Description
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
USP
Riboflavin, United States Pharmacopeia (USP) Reference Standard
Riboflavin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Supelco
Riboflavin (B2), analytical standard
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Riboflavin for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder