Merck
  • Effect of supraspinatus tendon injury on supraspinatus and infraspinatus muscle passive tension and associated biochemistry.
Effect of supraspinatus tendon injury on supraspinatus and infraspinatus muscle passive tension and associated biochemistry.
The Journal of bone and joint surgery. American volume (2014-10-17)
Morgan D Silldorff, Alexander D Choo, Anthony J Choi, Evie Lin, J Austin Carr, Richard L Lieber, John G Lane, Samuel R Ward
ABSTRACT
Injury to the supraspinatus and infraspinatus tendons and the associated atrophic changes to the muscle remain a common clinical problem. Specifically, increased muscle stiffness has been implicated in failure of the repair and poor functional outcomes. We present a comparison of the passive mechanical properties and associated biochemical studies from patients with and without torn supraspinatus tendons. Muscle biopsy samples (n = 40) were obtained from twenty patients undergoing arthroscopic shoulder surgery. Passive mechanical tests of both individual fibers and fiber bundles as well as analysis of titin molecular weight and collagen content were performed. At the fiber-bundle level, a significant increase in passive modulus was observed between intact supraspinatus samples (mean [and standard error], 237.41 ± 59.78 kPa) and torn supraspinatus samples (515.74 ± 65.48 kPa) (p < 0.05), a finding that was not observed at the single fiber level. Within the torn samples, elastic moduli in the supraspinatus were greater than in the infraspinatus at both the single fiber and the fiber-bundle level. There was a significant positive correlation between bundle elastic modulus and collagen content (r(2) = 0.465) in the supraspinatus muscle as well as a significant positive correlation between tear size and bundle elastic modulus (r(2) = 0.702) in the torn supraspinatus samples. Supraspinatus muscle passive tension increases in a tendon tear size-dependent manner after tendon injury. The increase in muscle stiffness appears to originate outside the muscle cell, in the extracellular matrix. Muscle stiffness after rotator cuff tendon injury is more severe with large tears. This finding supports the concept of early intervention, when tendon tears are smaller, and interventions targeting the extracellular matrix.
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder