Merck
  • Palladium charcoal-catalyzed Suzuki-Miyaura coupling to obtain arylpyridines and arylquinolines.
Palladium charcoal-catalyzed Suzuki-Miyaura coupling to obtain arylpyridines and arylquinolines.
The Journal of organic chemistry (2003-11-25)
Tsuyoshi Tagata, Mayumi Nishida
ABSTRACT
A phosphine ligand, such as PPh3 or 2-(dicyclohexylphosphino)biphenyl, is essential for the Pd/C-catalyzed Suzuki-Miyaura coupling of halopyridines and haloquinolines, although it has been reported that the reaction of phenyl chlorides can be catalyzed by nonprereduced Pd/C without any additives. In the reactions of bromopyridines, bromoquinolines, 2-chloropyridines, and 2-chloroquinolines, PPh3 was effective enough to provide coupling products in good yields. However, in the reactions of 3-chloropyridine, 4-chloropyridine, and 6-chloroquinoline, sterically hindered 2-(dicyclohexylphosphino)biphenyl was indispensable as a ligand.