Merck
  • Mechanistic insights on chaotropic interactions of liophilic ions with basic pharmaceuticals in polar ionic mode liquid chromatography.
Mechanistic insights on chaotropic interactions of liophilic ions with basic pharmaceuticals in polar ionic mode liquid chromatography.
Journal of chromatography. A (2014-10-15)
Edmond Sanganyado, Zhijiang Lu, Jay Gan
ABSTRACT
We report for the first time the effect of liophilic mobile phase additives on the mechanism of chiral recognition of basic chiral pharmaceutical on a vancomycin based chiral stationary phase (CSP). Using methanol as the mobile phase and 0.005% formic acid as pH modifier, we evaluated the effect of different concentrations of three types of liophilic anions, formate (HCOO(-)), nitrate (NO3(-)), and acetate (CH3COO(-)), on enantioresolution (Rs), enantioselectivity (α) and retention factor (k) of enantiomers of fluoxetine and atenolol. The effect of liophilic ion types on k followed the Hofmeister series: CH3COO(-)>HCOO(-)>NO3(-). Increasing concentration from 4 to 20mM resulted in decreases in Rs and k in accordance to hydrophobicity of the liophilic anion. The effect of temperature or mobile phase composition on enantioseparation was determined at 13-40°C. For both analytes, standard changes in enthalpy (ΔH°) and entropy (ΔS°) calculated using van't Hoff plots (lnk against 1/T) to varied from -4.99 to -0.63 kJ/mol and -11.82 to 9.47 J/mol, respectively. The van't Hoff plots showed elution order of the enantiomers of each analyte did not reverse in the temperature range studied. Chiral recognition of the enantiomers of atenolol and fluoxetine in the presence of liophilic ions was enthalpy driven.
Product Description
Supelco
Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
USP
Atenolol, United States Pharmacopeia (USP) Reference Standard
Atenolol, European Pharmacopoeia (EP) Reference Standard
Supelco
Acetic acid, analytical standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Triethylamine, ≥99.5%