Merck
  • Telomerase Cajal body protein 1 depletion inhibits telomerase trafficking to telomeres and induces G
Telomerase Cajal body protein 1 depletion inhibits telomerase trafficking to telomeres and induces G
Oncology letters (2014-08-15)
Ping Yuan, Zhitian Wang, Wang Lv, Hui Pan, Yunhai Yang, Xiaoshuai Yuan, Jian Hu
ABSTRACT
Telomerase Cajal body protein 1 (TCAB1) is a telomerase holoenzyme, which is markedly enriched in Cajal bodies (CBs) and facilitates the recruitment of telomerase to CBs in the S phase of the cell cycle. This recruitment is dependent on TCAB1 binding to a telomerase RNA component. The majority of cancer cells are able to grow indefinitely due to telomerase and its mechanism of trafficking to telomeres. In the present study, a certain level of TCAB1 expression in A549 human lung cells was identified and TCAB1 knockdown exhibited a potent antiproliferative effect on these cells, which was coupled with a decrease in the cell density and activity of the cellular enzymes. In addition, TCAB1-depletion was demonstrated to inhibit telomerase trafficking to telomeres in the A549 cells, leading to subsequent G
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Fluorescein, suitable for fluorescence, free acid
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Wdr79
Sigma-Aldrich
MISSION® esiRNA, targeting human WRAP53
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Fluorescein, European Pharmacopoeia (EP) Reference Standard