Merck
  • Gemcitabine diphosphate choline is a major metabolite linked to the Kennedy pathway in pancreatic cancer models in vivo.
Gemcitabine diphosphate choline is a major metabolite linked to the Kennedy pathway in pancreatic cancer models in vivo.
British journal of cancer (2014-05-31)
T E Bapiro, K K Frese, A Courtin, J L Bramhall, B Madhu, N Cook, A Neesse, J R Griffiths, D A Tuveson, D I Jodrell, F M Richards
ABSTRACT
The modest benefits of gemcitabine (dFdC) therapy in patients with pancreatic ductal adenocarcinoma (PDAC) are well documented, with drug delivery and metabolic lability cited as important contributing factors. We have used a mouse model of PDAC: KRAS(G12D); p53(R172H); pdx-Cre (KPC) that recapitulates the human disease to study dFdC intra-tumoural metabolism. LC-MS/MS and NMR were used to measure drug and physiological analytes. Cytotoxicity was assessed by the Sulphorhodamine B assay. In KPC tumour tissue, we identified a new, Kennedy pathway-linked dFdC metabolite (gemcitabine diphosphate choline (GdPC)) present at equimolar amounts to its precursor, the accepted active metabolite gemcitabine triphosphate (dFdCTP). Utilising additional subcutaneous PDAC tumour models, we demonstrated an inverse correlation between GdPC/dFdCTP ratios and cytidine triphosphate (CTP). In tumour homogenates in vitro, CTP inhibited GdPC formation from dFdCTP, indicating competition between CTP and dFdCTP for CTP:phosphocholine cytidylyltransferase (CCT). As the structure of GdPC precludes entry into cells, potential cytotoxicity was assessed by stimulating CCT activity using linoleate in KPC cells in vitro, leading to increased GdPC concentration and synergistic growth inhibition after dFdC addition. GdPC is an important element of the intra-tumoural dFdC metabolic pathway in vivo.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, sodium pyruvate, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose and sodium bicarbonate, without L-glutamine and sodium pyruvate, liquid, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Gemcitabine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Linoleic acid sodium salt, ≥97% (GC)
Gemcitabine hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Gemcitabine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Cytidine, BioReagent, suitable for cell culture, powder, ≥99%
Sigma-Aldrich
Cytidine, 99%
Sigma-Aldrich
Cytidine 5′-diphosphocholine sodium salt dihydrate, ~98%, from yeast, solid
Sigma-Aldrich
6-Diazo-5-oxo-L-norleucine, powder