Merck
  • Intracellular MMP-2 activity in skeletal muscle is associated with type II fibers.
Intracellular MMP-2 activity in skeletal muscle is associated with type II fibers.
Journal of cellular physiology (2014-06-07)
Elin Hadler-Olsen, Ann Iren Solli, Anne Hafstad, Jan-Olof Winberg, Lars Uhlin-Hansen
ABSTRACT
Matrix metalloproteinase 2 (MMP-2) is a proteolytic enzyme implicated in motility, differentiation, and regeneration of skeletal muscle fibers through processing of extracellular substrates. Although MMP-2 has been found to be localized intracellularly in cardiomyocytes where the enzyme is thought to contribute to post-ischemic loss of contractility, little is known about intracellular MMP-2 activity in skeletal muscle fibers. In the present study we demonstrate intracellular MMP-2 in normal skeletal muscle by immunohistochemical staining. Immunogold electron microscopic analyses indicated that the enzyme was concentrated in Z-lines of the sarcomers, in the nuclear membrane, and in mitochondria. By use of in situ zymography, we found that gelatinolytic activity in muscle fibers was co-localized with immunofluorecent staining for MMP-2. Staining for MMP-9, the other member of the gelatinase group of the MMPs, was negative. The broad-spectrum metalloprotease inhibitor EDTA and the selective gelatinase inhibitor CTT2, but not the cysteine inhibitor E64, strongly reduced the gelatinolytic activity. The intracellular gelatinolytic activity was much more prominent in fast twitch type II fibers than in slow twitch type I fibers, and there was a decrease in intracellular gelatinolytic activity and MMP-2 expression in muscles from mice exposed to high intensity interval training. Together our results indicate that MMP-2 is part of the intracellular proteolytic network in normal skeletal muscle, especially in fast twitch type II fibers. Further, the results suggest that intracellular MMP-2 in skeletal muscle fibers is active during normal homeostasis, and affected by the level of physical activity.
Product Description
Sodium calcium edetate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, Molecular Biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sigma-Aldrich
E-64, protease inhibitor
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid calcium disodium salt hydrate
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, 98.5-101.5%
Sigma-Aldrich
L-Leucine, reagent grade, ≥98% (HPLC)
Supelco
L-Leucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Leucine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Sigma-Aldrich
L-Leucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
L-Leucine, 99%, FG
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
L-Leucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Supelco
Ethylenediaminetetraacetic acid disodium salt dihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Edetate disodium, United States Pharmacopeia (USP) Reference Standard