Merck
  • Gender differences in pharmacokinetics of a combination tablet of niacin extended-release/simvastatin in healthy Chinese volunteers.
Gender differences in pharmacokinetics of a combination tablet of niacin extended-release/simvastatin in healthy Chinese volunteers.
European journal of drug metabolism and pharmacokinetics (2013-12-19)
Xiao-lin Wang, Man Liu, Man Yang, Ya-nan Zhang, Dan Zhang, Li-na Zhang, Jing Han, Hui-chen Liu
ABSTRACT
The gender differences in pharmacokinetics of a combination tablet of niacin extended-release/simvastatin were evaluated in healthy Chinese volunteers. Thirty-six healthy male and female volunteers were enrolled in the study receiving a single oral dose of niacin extended-release/simvastatin 1,000/20 mg. The results indicated that the systemic exposure of simvastatin hydroxy acid and the total urine excretion of niacin were significantly higher for females compared with those for males, and the T max of niacin in plasma was significantly shorter for males than that for females. There were no significant differences in the systemic exposure of simvastatin, niacin, and NUA in plasma between males and females.
Product Description
USP
Simvastatin, United States Pharmacopeia (USP) Reference Standard
Simvastatin, European Pharmacopoeia (EP) Reference Standard
Supelco
Simvastatin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Supelco
Simvastatin, analytical standard
Sigma-Aldrich
Nicotinic acid sodium salt, 98%
Supelco
Nicotinic acid, analytical standard
Sigma-Aldrich
Nicotinic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Nicotinic acid, ≥98%
Sigma-Aldrich
Nicotinic acid, meets USP testing specifications
Sigma-Aldrich
Nicotinic acid, ≥99.5% (HPLC)
Supelco
Niacin (Nicotinic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
USP
Niacin, United States Pharmacopeia (USP) Reference Standard
Nicotinic acid, European Pharmacopoeia (EP) Reference Standard